Skip to Content
Merck
All Photos(1)

Key Documents

N-006

Supelco

Normorphine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17NO3
CAS Number:
Molecular Weight:
271.31
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

O[C@H]1C=CC2[C@H]3Cc4ccc(O)c5O[C@@H]1[C@]2(CCN3)c45

InChI

1S/C16H17NO3/c18-11-3-1-8-7-10-9-2-4-12(19)15-16(9,5-6-17-10)13(8)14(11)20-15/h1-4,9-10,12,15,17-19H,5-7H2/t9?,10-,12+,15+,16+/m1/s1

InChI key

ONBWJWYUHXVEJS-YTPBOXJFSA-N

General description

Normorphine is a primary urinary metabolite of the opiate analgesic morphine. This Snap-N-Spike® reference solution is applicable for use in LC/MS or GC/MS applications such as pain prescription monitoring, urine drug testing, clinical toxicology, or forensic analysis. Morphine, used to relieve both acute and chronic severe pain is known under trade names MS Contin®, Kadian®, Oramorph®, and Roxanol.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Kadian is a registered trademark of Actavis Elizabeth LLC
MS Contin is a registered trademark of Purdue Frederick Company
Oramorph is a registered trademark of AAIPharma, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Pawula et al.
Journal of pharmaceutical and biomedical analysis, 11(4-5), 401-406 (1993-04-01)
A new, simple and rapid extraction procedure coupled with a combined coulometric-fluorescence HPLC assay is described for the simultaneous determination of morphine (M) and morphine-3-glucuronide (M3G), morphine-6-glucuronide (M6G), and normorphine (NM) in plasma. The effect of concentration and pH of
J Hasselström et al.
Pharmacology & toxicology, 79(1), 40-46 (1996-07-01)
Morphine, morphine-6-glucuronide and normorphine were administered to male Sprague-Dawley-rats. Analgesic effect was estimated with the hot plate and spinal nociceptive reflex depression. After intraperitoneal administration the molar potency ratio of morphine-6-glucuronide/morphine was 1.7 estimated by the paw lick latency on
A A Coutts et al.
Canadian journal of physiology and pharmacology, 76(3), 340-346 (1998-07-23)
Cannabinoid receptor agonists inhibit electrically evoked isometric contractions of the myenteric plexus--longitudinal muscle preparation of the guinea-pig small intestine (MPLM), probably by reducing release of acetylcholine (ACh) through the activation of prejunctional CB1 receptors. As CB1 receptors are thought to
O Dale et al.
European journal of clinical pharmacology, 63(9), 837-842 (2007-07-11)
To compare the time course of morphine and metabolite concentrations in serum and cerebrospinal fluid (CSF) after intravenous and intramuscular administration after surgery. This was a randomized double-blind, double-dummy study in patients who had undergone hip replacement surgery. Morphine (M
Yi-Hui Lin et al.
Electrophoresis, 29(11), 2340-2347 (2008-04-26)
A cation-selective exhaustive injection and sweeping micellar EKC (CSEI-Sweep-MEKC) was established to analyze morphine and its four metabolites, including codeine, normorphine (NM), morphine-3-glucuronide (M3G), and morphine-6-glucuronide (M6G). After SPE, the urine samples were analyzed by this CE method. The phosphate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service