Skip to Content
Merck
All Photos(1)

Documents

M-137

Supelco

Mycophenolic acid-d3 solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17O6D3
CAS Number:
Molecular Weight:
323.36
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

C/C(CCC(O)=O)=C\CC1=C(O)C2=C(COC2=O)C(C)=C1OC([2H])([2H])[2H]

InChI

1S/C17H20O6/c1-9(5-7-13(18)19)4-6-11-15(20)14-12(8-23-17(14)21)10(2)16(11)22-3/h4,20H,5-8H2,1-3H3,(H,18,19)/b9-4+/i3D3

InChI key

HPNSFSBZBAHARI-HZIIEIAOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A Certified Spiking Solution® and internal standard for use in the quantitation of mycophenolic acid levels in serum or whole blood by LCMS. Assayed in therapeutic drug monitoring to ensure patients remain within the drug′s therapeutic range, mycophenolic acid is an immunosuppressant drug and the active metabolite of the prodrug mycophenolate mofetil.

Application


  • Ultra Performance Liquid Chromatography in Pharmacokinetics: A study utilizes ultra-performance liquid chromatography tandem mass spectrometry (UPLC/MS/MS) to determine mycophenolic acid in human plasma, enhancing drug monitoring in clinical settings. This technique offers high sensitivity and specificity, critical for therapeutic drug monitoring of immunosuppressants used in transplant medicine (Upadhyay et al., 2014).

  • Comparative Methods for Immunosuppressant Monitoring: This research compares two rapid ultra-performance liquid chromatography/tandem mass spectrometry methods for therapeutic drug monitoring of immunosuppressants, including mycophenolic acid, against traditional immunoassay techniques. The study highlights the precision and faster turnaround times of UPLC/MS/MS methods, offering significant improvements in patient care management for those undergoing immunosuppressive therapy (Tszyrsznic et al., 2013).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Safraaz Mahamed et al.
The Journal of physiology, 586(8), 2171-2181 (2008-02-23)
Long-term facilitation (LTF) is a form of respiratory neuroplasticity frequently induced by acute intermittent isocapnic hypoxia (AIH, three 5 min isocapnic hypoxic episodes). Although repetitive apnoeas are a frequent natural occurrence producing brief (< 30 s) episodes of hypoxia and
Miguel Toro-Sazo et al.
PloS one, 14(1), e0209804-e0209804 (2019-01-11)
The last fifteen years have seen the emergence and overflow into the drug scene of "superpotent" N-benzylated phenethylamines belonging to the "NBOMe" series, accompanied by numerous research articles. Although N-benzyl substitution of 5-methoxytryptamine is known to increase its affinity and
John D McCorvy et al.
Nature structural & molecular biology, 25(9), 787-796 (2018-08-22)
Serotonin (5-hydroxytryptamine; 5-HT) receptors modulate a variety of physiological processes ranging from perception, cognition and emotion to vascular and smooth muscle contraction, platelet aggregation, gastrointestinal function and reproduction. Drugs that interact with 5-HT receptors effectively treat diseases as diverse as
L L Hernandez et al.
Journal of dairy science, 91(5), 1834-1844 (2008-04-19)
Serotonin (5-HT), a neurotransmitter synthesized from tryptophan, has been proposed as a feedback inhibitor of lactation. We determined that the gene coding for tryptophan hydroxylase 1, the rate-limiting enzyme for 5-HT synthesis, is expressed in bovine mammary epithelial cells in
Lingyan Xing et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(44), 14794-14808 (2015-11-06)
Modulation of connectivity formation in the developing brain in response to external stimuli is poorly understood. Here, we show that the raphe nucleus and its serotonergic projections regulate pathfinding of commissural axons in zebrafish. We found that the raphe neurons

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service