Skip to Content
Merck
All Photos(1)

Key Documents

M-072

Supelco

Metformin hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H12ClN5
CAS Number:
Molecular Weight:
165.62
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

Cl.CN(C)C(=N)NC(N)=N

InChI

1S/C4H11N5.ClH/c1-9(2)4(7)8-3(5)6;/h1-2H3,(H5,5,6,7,8);1H

InChI key

OETHQSJEHLVLGH-UHFFFAOYSA-N

Gene Information

General description

A Snap-N-Spike® reference solution applicable to use in clinical toxicology by LC-MS/MS or GC/MS. Metformin is an antidiabetic drug which functions by suppressing liver production of glucose. This biguanide analog is used specifically for treatment of type 2 diabetes.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Amador-Licona et al.
Archives of medical research, 31(6), 571-575 (2001-03-21)
Renal hyperfiltration and albuminuria have a deleterious effect on kidney function. Therefore, we studied the effect of metformin on blood pressure, renal hemodynamics, and microalbuminuria in type 2 diabetic patients. A clinical trial was designed in type 2 diabetic patients
Nicolas Musi et al.
Diabetes, 51(7), 2074-2081 (2002-06-28)
Metformin is an effective hypoglycemic drug that lowers blood glucose concentrations by decreasing hepatic glucose production and increasing glucose disposal in skeletal muscle; however, the molecular site of metformin action is not well understood. AMP-activated protein kinase (AMPK) activity increases
L C Morin-Papunen et al.
The Journal of clinical endocrinology and metabolism, 85(9), 3161-3168 (2000-09-22)
Metformin, a biguanide antihyperglycemic drug, has been shown to improve ovarian function and glucose metabolism in women with polycystic ovary syndrome (PCOS), but results concerning its effects on insulin sensitivity are controversial. Oral contraceptive pills are commonly used in the
J De Jager et al.
Journal of internal medicine, 257(1), 100-109 (2004-12-21)
The UK Prospective Diabetes Study (UKPDS) showed that treatment with metformin decreases macrovascular morbidity and mortality independent of glycaemic control. We hypothesized that metformin may achieve this by improving endothelial function and chronic, low-grade inflammation. Data on this issue are
Yingwu Wang et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 808(2), 215-219 (2004-07-21)
A rapid and sensitive liquid chromatography-tandem mass spectrometric (LC-MS-MS) method for the determination of metformin in human plasma using phenformin as internal standard has been developed and validated. Sample preparation of plasma involved acidification with acetic acid, deproteination with acetonitrile

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service