Skip to Content
Merck
All Photos(1)

Key Documents

D-079

Supelco

Demoxepam solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11ClN2O2
CAS Number:
Molecular Weight:
286.71
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

ClC1=CC=C(NC(C[N+]([O-])=C2C3=CC=CC=C3)=O)C2=C1

InChI

1S/C15H11ClN2O2/c16-11-6-7-13-12(8-11)15(10-4-2-1-3-5-10)18(20)9-14(19)17-13/h1-8H,9H2,(H,17,19)

InChI key

GGRWZBVSUZZMKS-UHFFFAOYSA-N

General description

A benzodiazepine drug and anticonvulsant, demoxepam is also a primary metabolite of chlordiazepoxide, marketed as Klopoxid or Librax® for the treatment of anxiety and muscle spasms or pain.

Application


  • Pharmacokinetic and Pharmacodynamic Analysis: A study discussed the rapid hydrolysis of chlordiazepoxide to Demoxepam, emphasizing the importance of understanding metabolic pathways in benzodiazepine analogues research. This knowledge aids in the design of neuropharmacology experiments and the development of GABA receptor agonist studies (Vinkers et al., 2010).

  • Neuropharmacological Studies: Research utilizing Demoxepam has explored its role as a benzodiazepine receptor ligand, which provides insight into its function as a neuropharmacology research chemical. These studies contribute to our understanding of Demoxepam′s effects on anxiety and its potential therapeutic applications (Lister et al., 1984).

  • High-Performance Liquid Chromatography (HPLC) Applications: The application of HPLC in the determination of 1,4-benzodiazepines and their metabolites, including Demoxepam, highlights its utility in the precise measurement of these compounds in plasma. This is crucial for biochemical assays and enhances the research reagent′s reliability in pharmaceutical industry studies (Skellern et al., 1978).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Librax is a registered trademark of Valeant Pharmaceuticals North America LLC
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moustapha Ouédraogo et al.
Drug and chemical toxicology, 32(4), 417-423 (2009-10-02)
The cytotoxicity of oxaziridines photogenerated after irradiation of chlordiazepoxide (CDZ) and its metabolites was investigated in vitro by a MTT assay on P388 leukemia and B16 melanoma cell lines and compared with that of the anticancer drug, melphalan. For the
A G Davidson
Journal of pharmaceutical sciences, 73(1), 55-58 (1984-01-01)
Rapid difference spectrophotometric methods for chlordiazepoxide and demoxepam in chlordiazepoxide formulations are described which overcome the nonspecificity of the official spectrophotometric assays. The procedures are based on the measurement of the difference absorbance at 269 nm of equimolar solutions of
U A Brinkman et al.
Journal of chromatography, 217, 463-471 (1981-11-06)
Upon irradiation with short-wavelength UV light the tranquillizer demoxepam is converted into a highly fluorescent product. This reaction serves as a sensitive and selective means of detection of low levels of demoxepam in e.g., serum after separation by means of
H Essien et al.
Journal of chromatography. B, Biomedical applications, 683(2), 199-208 (1996-08-30)
The identification of the metabolite demoxepam in human urine establishes that chlordiazepoxide, a common benzodiazepine, has been administered. Like N-oxide metabolites of other drugs, demoxepam cannot be detected by gas chromatography-mass spectrometry (GC-MS), due to thermal decomposition, and the product
T J Good et al.
Journal of chromatographic science, 19(11), 562-566 (1981-11-01)
A simple, rapid method for the determination of the benzodiazepines, diazepam and chlordiazepoxide, and their metabolites by high performance liquid chromatography (HPLC) has been developed. The procedure is applicable to the assay of other similar drugs in biological fluids. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service