Skip to Content
Merck
All Photos(1)

Key Documents

D-058

Supelco

O-Desmethyl-cis-tramadol-D6 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15D6H17NO2 · HCl
Molecular Weight:
291.85
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

OC1=CC([C@@]2(O)CCCC[C@@H]2CN(C([2H])([2H])[2H])C([2H])([2H])[2H])=CC=C1.Cl

InChI

1S/C15H23NO2.ClH/c1-16(2)11-13-6-3-4-9-15(13,18)12-7-5-8-14(17)10-12;/h5,7-8,10,13,17-18H,3-4,6,9,11H2,1-2H3;1H/t13-,15+;/m1./s1/i1D3,2D3;

InChI key

IRGWVAWLHXDKIX-YATKDSQESA-N

General description

O-Desmethyl-cis-tramadol is the primary, active metabolite of tramadol, marketed as the opioid analgesic Ultram® or Tramal, for the treatment of moderate to moderately severe pain. This new stable-labeled internal standard is suitable for the quantitation of O-Desmethyl-cis-tramadol in biological samples using GCMS or LCMS with numerous applications including forensic or clinical toxicology analysis, urine drug testing, or isotope dilution methods.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Ultram is a registered trademark of Johnson & Johnson

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service