Skip to Content
Merck
All Photos(1)

Key Documents

880730P

Avanti

18:1 PEG1000 PE

Avanti Research - A Croda Brand 880730P, powder

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-1000] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 2 × 100 mg (880730P-200mg)
pkg of 1 × 25 mg (880730P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880730P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+]

InChI

1S/C87H168NO32P.H3N/c1-4-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-85(89)117-82-84(120-86(90)35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-5-2)83-119-121(92,93)118-37-36-88-87(91)116-81-80-115-79-78-114-77-76-113-75-74-112-73-72-111-71-70-110-69-68-109-67-6

InChI key

GAPHFOOXSGWGKG-QOYAZRIPSA-N

Application

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-1000] (18:1 PEG1000 PE) is suitable for fluorescent PEG-grafted liposomes/fluorescent stealth liposomes or liposomes.

Biochem/physiol Actions

Polyethylene glycol (PEG) conjugated phospholipids are known to protect the steric nature of liposomes. Hence, they are commonly recommended for use in liposome formulation.

Packaging

5 mL Clear Glass Sealed Ampule (880730P-200mg)
5 mL Clear Glass Sealed Ampule (880730P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surface-grafted polyethylene glycol conformation impacts the transport of PEG-functionalized liposomes through a tumour extracellular matrix model
Labouta HI, et al.
Royal Society of Chemistry Advances, 8(14), 7697-7708 (2018)
Have Aliu et al.
International journal of nanomedicine, 12, 8377-8388 (2017-12-05)
Immunotherapy by sublingual administration of allergens provides high patient compliance and has emerged as an alternative to subcutaneous immunotherapy for the treatment of IgE-associated allergic diseases. However, sublingual immunotherapy (SLIT) can cause adverse events. Development of allergen delivery systems enabling
G F Peacock et al.
Drug delivery, 10(1), 29-34 (2003-01-30)
A new cholesterol-carborane conjugate (BCH) has been synthesized as a potential targeting agent for boron neutron capture therapy (BNCT) of cancers. The compound is extremely water insoluble and was formulated in two liposomal formulations to determine if the compound could
Maria C Sandström et al.
Langmuir : the ACS journal of surfaces and colloids, 23(8), 4192-4198 (2007-03-09)
Polyethylene glycol (PEG)-conjugated lipids are commonly employed for steric stabilization of liposomes. When added in high concentrations PEG-lipids induce formation of mixed micelles, and depending on the lipid composition of the sample, these may adapt either a discoidal or a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service