Skip to Content
Merck
All Photos(1)

Key Documents

870314C

Avanti

18:1 DGS

Avanti Research - A Croda Brand 870314C

Synonym(s):

1,2-dioleoyl-sn-glycero-3-succinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H76O8
CAS Number:
Molecular Weight:
721.06
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 2.5 mL (870314C-25mg)
pkg of 2 × 4 mL (870314C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870314C

concentration

10 mg/mL (870314C-25mg)
25 mg/mL (870314C-200mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COC(CCC(O)=O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C43H76O8/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-41(46)49-37-39(38-50-42(47)36-35-40(44)45)51-43(48)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20,39H,3-16,21-38H2,1-2H3,(H,44,45)/b19-17-,20-18-/t39-/m1/s1

InChI key

VZGIZLLBNVAMQT-NYVOMTAGSA-N

General description

18:1 DGS or 1,2-dioleoyl-sn-glycero-3-succinate (DOSG) is a non-phospholipid. This lipid contains double oleoyl chains and all of this chains form stable fluidic bilayers.

Application

1,2-dioleoyl-sn-glycero-3-succinate (DOSG) or 18:1 DGS has been used in liposome preparation.

Packaging

5 mL Clear Glass Sealed Ampule (870314C-200mg)
5 mL Clear Glass Sealed Ampule (870314C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipid-dependent gating of a voltage-gated potassium channel
Zheng H, et al.
Nature Communications, 2(3), 250-250 (2011)
Delivery of superoxide dismutase to pulmonary epithelium via pH-sensitive liposomes
Briscoe P, et al.
American Journal of Physiology. Lung Cellular and Molecular Physiology, 268(3), L374-L380 (1995)
Sungsoo Lee et al.
Journal of visualized experiments : JoVE, (77), e50436-e50436 (2013-07-31)
To study the lipid-protein interaction in a reductionistic fashion, it is necessary to incorporate the membrane proteins into membranes of well-defined lipid composition. We are studying the lipid-dependent gating effects in a prototype voltage-gated potassium (Kv) channel, and have worked

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service