Skip to Content
Merck
All Photos(1)

Key Documents

870013P

Avanti

16:0 MPB PE

Avanti Research - A Croda Brand 870013P, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-[4-(p-maleimidophenyl)butyramide] (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H84N2O11PNa
CAS Number:
Molecular Weight:
955.18
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 100 mg (870013P-100mg)
pkg of 1 × 25 mg (870013P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870013P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(CCCC(C=C1)=CC=C1N(C(C=C2)=O)C2=O)=O)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C51H85N2O11P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-32-50(57)61-42-46(64-51(58)33-28-26-24-22-20-18-16-14-12-10-8-6-4-2)43-63-65(59,60)62-41-40-52-47(54)31-29-30-44-34-36-45(37-35-44)53-48(55)38-39-49(53)56;/h34-39,46H,3-33,40-43H2,1-2H3,(H,52,54)(H,59,60);/q;+1/p-1/t46-;/m1./s1

InChI key

REKODAHLWDSEBQ-FYQWNKFKSA-M

General description

16:0 maleimidophenyl butyramide (MPB) phosphoethanolamine (PE) also known as, DPPE-maleimide contains maleimide, which is a thiol-reactive moiety and a good electrophile. MPB PE is implicated in liposome preparation.

Application

16:0 maleimidophenyl butyramide (MPB) phosphoethanolamine (PE) is suitable for use:
  • for preparation of maleimide-functionalized microbubbles
  • for lipidation of ligand binding site-1 (LBS-1) peptide and GABAA receptor-associated protein (GABARAP) involved in liposome and autophagosome formation, respectively
  • for preparation of ligand-conjugated acoustic perfluorocarbon emulsion nanoparticles

Packaging

5 mL Amber Glass Screw Cap Vial (870013P-100mg)
5 mL Amber Glass Screw Cap Vial (870013P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grafting of abciximab to a microbubble-based ultrasound contrast agent for targeting to platelets expressing GP IIb/IIIa--characterization and in vitro testing
Della MA, et al.
European Journal of Pharmaceutics and Biopharmaceutics, 68(3), 555-564 (2008)
Preparation of a functional GABARAP--lipid conjugate in nanodiscs and its investigation by solution NMR spectroscopy
Ma P, et al.
Chembiochem, 11(14), 1967-1970 (2010)
Novel MRI contrast agent for molecular imaging of fibrin: implications for detecting vulnerable plaques
Flacke S, et al
Circulation, 104(11), 1280-1285 (2001)
Liposomes as tools in basic research and industry (1994)
Structural basis for thrombin activation of a protease-activated receptor: inhibition of intramolecular liganding
Seeley S, et al.
Chemistry & Biology, 10(11), 1033-1041 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service