Skip to Content
Merck
All Photos(1)

Key Documents

860689P

Avanti

C15 Ceramide-1-Phosphate-d7 (d18:1-d7/15:0)

Avanti Research - A Croda Brand (860689P), powder

Synonym(s):

N-pentadecanoyl-ceramide-1-phosphate-d7 (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H62D7N2O6P
CAS Number:
Molecular Weight:
627.93
UNSPSC Code:
12352100
NACRES:
NA.12

Assay

99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (860689P-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand (860689P)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCC([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])(O)[C@@]([H])(NC(CCCCCCCCCCCCCC)=O)COP([O-])(O)=O.[NH4+]

InChI key

GXIXNMIIJBAQTF-QHKXSBLVSA-N

General description

C15 Ceramide-1-Phosphate-d7 (d18:1-d7/15:0) is a deuterated derivative of C15 Ceramide-1-Phosphate. Ceramide-1-Phosphate (C1P) is a bioactive sphingolipid. It is a metabolite of ceramide and is obtained by the phosphorylation of ceramide by ceramide kinase.

Biochem/physiol Actions

Ceramide-1-Phosphate (C1P) plays an important role in regulating apoptosis and cell proliferation. It is also involved in phagocytosis and is an important factor in the modulation of macrophage chemotaxis. C1P possesses pro-inflammatory activity.

Packaging

5 mL Amber Glass Screw Cap Vial

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of ceramide-1-phosphate in biological functions
Sphingolipids: Basic Science and Drug Development, 153-166 (2013)
Ceramide and ceramide 1-phosphate in health and disease
Arana L, et al.
Lipids in Health and Disease, 9(1), 15-15 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service