Skip to Content
Merck
All Photos(1)

Key Documents

860652P

Avanti

C8 Ceramide-1-Phosphate (d17:1/8:0)

Avanti Research - A Croda Brand 860652P, powder

Synonym(s):

N-octanoyl-ceramide-1-phosphate (C17 base) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H53N2O6P
CAS Number:
Molecular Weight:
508.67
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860652P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860652P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCC/C=C/[C@@]([H])(O)[C@@]([H])(NC(CCCCCCC)=O)COP([O-])(O)=O.[NH4+]

InChI key

QZINIJMZYDYICQ-ITEYRTANSA-N

General description

C8 Ceramide-1-Phosphate (d17:1/8:0) is a novel bioactive sphingolipid. It is formed by the phosphorylation of C8-ceramide or N-octanoyl-D-erythro-sphingosine (C17 base) catalyzed by ceramide kinase (CERK). C1P is a sphingolipid analog of phosphatidate.

Biochem/physiol Actions

Ceramide 1-phosphate (C1P) acts as a mitogenic agent. It exhibits antiapoptotic and prosurvival properties. C1P stimulates inflammatory response and facilitates phagocytosis. It also has an ability to negatively regulate tumor necrosis factor-α (TNFα) production induced by lipopolysaccharide. C1P is reputed to be a second lipid messenger of agonist-stimulated sphingomyelin metabolism.

Packaging

5 mL Amber Glass Screw Cap Vial (860652P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tack-Joong Kim et al.
Experimental cell research, 317(14), 2041-2051 (2011-06-01)
Ceramide 1-phosphate (C1P) is a novel bioactive sphingolipid formed by ceramide kinase (CERK)-catalyzed phosphorylation of ceramide. It has been implicated in the regulation of such vital pathophysiological functions as phagocytosis and inflammation, but there have been no reports ascribing a
Lide Arana et al.
Lipids in health and disease, 9, 15-15 (2010-02-09)
Sphingolipids are essential components of cell membranes, and many of them regulate vital cell functions. In particular, ceramide plays crucial roles in cell signaling processes. Two major actions of ceramides are the promotion of cell cycle arrest and the induction
Szczepan Józefowski et al.
Journal of immunology (Baltimore, Md. : 1950), 185(11), 6960-6973 (2010-11-03)
LPS is a constituent of cell walls of Gram-negative bacteria that, acting through the CD14/TLR4 receptor complex, causes strong proinflammatory activation of macrophages. In murine peritoneal macrophages and J774 cells, LPS at 1-2 ng/ml induced maximal TNF-α and MIP-2 release
Antonio Gómez-Muñoz et al.
Journal of lipid research, 45(1), 99-105 (2003-10-03)
It was reported previously that ceramide-1-phosphate (Cer-1-P) is mitogenic for fibroblasts (Gómez-Muñoz, A., P. A. Duffy, A. Martin, L. O'Brien, H-S. Byun, R. Bittman, and D. N. Brindley. 1995. Mol. Pharmacol. 47: 883-889; Gómez-Muñoz, A., L. M. Frago, L. Alvarez
A Gomez-Muñoz et al.
Molecular pharmacology, 47(5), 833-839 (1995-05-01)
Ceramide and ceramide-1-phosphate are sphingolipid analogues of diacylglycerol and phosphatidate, respectively, and they are putative second messengers of agonist-stimulated sphingomyelin metabolism. The interactions of exogenous cell-permeable ceramides and ceramide-1-phosphates in modifying DNA synthesis and signal transduction were investigated in Rat-1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service