Skip to Content
Merck
All Photos(1)

Documents

860600P

Avanti

Lyso SM (d18:1)

Avanti Research - A Croda Brand 860600P, powder

Synonym(s):

Sphingosylphosphorylcholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H49N2O5P
CAS Number:
Molecular Weight:
464.62
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (860600P-10mg)
pkg of 1 × 25 mg (860600P-25mg)
pkg of 1 × 5 mg (860600P-5mg)
pkg of 1 × 50 mg (860600P-50mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860600P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(N)COP([O-])(OCC[N+](C)(C)C)=O

InChI

1S/C23H49N2O5P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-23(26)22(24)21-30-31(27,28)29-20-19-25(2,3)4/h17-18,22-23,26H,5-16,19-21,24H2,1-4H3/b18-17+/t22-,23+/m0/s1

InChI key

JLVSPVFPBBFMBE-HXSWCURESA-N

General description

Lyso SM (d18:1), also known as sphingosylphosphorylcholine (SPC), is a bioactive lipid molecule. This lysophospholipid is present in normal blood plasma, ascites and various tissues.

Application

Lyso SM (d18:1) has been used:
  • to study its effect on the skeletal muscle ryanodine receptor (RyR1), in the presence and absence of calmodulin through single channel recordings in planar lipid bilayers
  • as an inhibitor of Ca2+ sensor calmodulin (CaM)
  • as a standard in circular dichroism (CD) spectroscopy to determine lipid selectivity of the lipid-peptide interactions

Biochem/physiol Actions

Lyso SM (d18:1) or sphingosylphosphorylcholine (SPC) is a lipid mediator that exhibits second messenger functions. It regulates ryanodine receptors (RyRs) and releases Ca2+ from the sarco/endoplasmic reticulum. Lyso SM (d18:1) acts as an endogenous inhibitor of ubiquitous Ca2+ sensor calmodulin (CaM).

Packaging

5 mL Amber Glass Screw Cap Vial (860600P-10mg)
5 mL Amber Glass Screw Cap Vial (860600P-25mg)
5 mL Amber Glass Screw Cap Vial (860600P-50mg)
5 mL Amber Glass Screw Cap Vial (860600P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sphingosylphosphorylcholine and lysophosphatidylcholine are ligands for the G protein-coupled receptor GPR4.
The Journal of biological chemistry, 280(52), 43280-43280 (2006-02-25)
Dissociation of Calmodulin-Target Peptide Complexes by the Lipid Mediator Sphingosylphosphorylcholine IMPLICATIONS IN CALCIUM SIGNALING
Kovacs E, et al.
The Journal of Biological Chemistry, 285(3), 1799-1808 (2010)
Sphingosylphosphorylcholine and lysophosphatidylcholine are ligands for the G protein-coupled receptor GPR4
Zhu k, et al.
The Journal of Biological Chemistry, 276(44), 41325-41335 (2001)
Dagmar Meyer zu Heringdorf et al.
Biochimica et biophysica acta, 1582(1-3), 178-189 (2002-06-19)
Compared to the lysophospholipid mediators, sphingosine-1-phosphate (S1P) and lysophosphatidic acid (LPA), little information is available regarding the molecular mechanisms of action, metabolism and physiological significance of the related sphingosylphosphorylcholine (SPC). S1P and LPA have recently been established as agonists at
Sphingosylphosphorylcholine-biological functions and mechanisms of action.
Meyer ZU, et al.
Biochimica et Biophysica Acta, 1582(1-3), 178-178 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service