Skip to Content
Merck
All Photos(1)

Key Documents

860599P

Avanti

C18:1 Ceramide-1-Phosphate (d18:1/18:1(9Z))

Avanti Research - A Croda Brand 860599P, powder

Synonym(s):

N-oleoyl-ceramide-1-phosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H73N2O6P
CAS Number:
Molecular Weight:
660.95
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860599P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860599P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCC/C=C\CCCCCCCC)=O)COP([O-])(O)=O.[NH4+]

InChI

1S/C36H70NO6P.H3N/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-36(39)37-34(33-43-44(40,41)42)35(38)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h17-18,29,31,34-35,38H,3-16,19-28,30,32-33H2,1-2H3,(H,37,39)(H2,40,41,42);1H3/b18-17-,31-29+;/t34-,35+;/m0./s1

InChI key

OLFFCEGYMCQDKZ-CFRDMXQMSA-N

General description

Ceramide-1-phosphate (C1P) is a ceramide-derived metabolite. It belongs to the class of bioactive lipid mediators.

Application

C18:1 Ceramide-1-Phosphate (d18:1/18:1(9Z)) is suitable for use in fluorescence analysis of phosphatidic acid-binding tetravalent peptide (PAB-TP) phospholipid specificity. It is also suitable for use in liposome cosedimentation assay to compare its affinity with α-synuclein-N.

Biochem/physiol Actions

Ceramide-1-phosphate (C1P) regulates inflammatory response and phagocytosis. It is also implicated in cell survival, cell growth and death.

Packaging

5 mL Amber Glass Screw Cap Vial (860599P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of alpha-synuclein N-terminal domain as a novel cellular phosphatidic acid sensor
Yamada H, et al.
The FEBS journal (2019)
Blood sphingolipidomics in healthy humans: impact of sample collection methodology
Hammad SM, et al.
Journal of Lipid Research, 51(10), 3074-3087 (2010)
Characterization of alpha-synuclein N-terminal domain as a novel cellular phosphatidic acid sensor
Yamada H, et al.
FEBS Journal (2019)
Development of a novel tetravalent synthetic peptide that binds to phosphatidic acid
Ogawa R, et al.
Testing, 10(7) (2015)
Development of a novel tetravalent synthetic peptide that binds to phosphatidic acid
Ogawa R, et al.
PLoS ONE, 10(7) (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service