Skip to Content
Merck
All Photos(1)

Key Documents

860576P

Avanti

C16 Lactosyl(β) Ceramide (d18:1/16:0)

Avanti Research - A Croda Brand 860576P, powder

Synonym(s):

D-lactosyl-β-1,1′ N-palmitoyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H87NO13
CAS Number:
Molecular Weight:
862.18
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (860576P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860576P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@@H]([C@H]1CO)O[C@H](O2)[C@H](O)[C@@H](O)[C@H]([C@H]2CO)O

InChI

1S/C46H87NO13/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-35(50)34(47-38(51)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)33-57-45-43(56)41(54)44(37(32-49)59-45)60-46-42(55)40(53)39(52)36(31-48)58-46/h27,29,34-37,39-46,48-50,52-56H,3-26,28,30-33H2,1-2H3,(H,47,51)

InChI key

HLIJNIKSBCIDGO-QKLMXXKVSA-N

General description

Lactosylceramide (LacCer) is a glycosphingolipid, which is found majorly in the nervous system and myelin sheath. It comprises a hydrophobic ceramide lipid and a hydrophilic sugar moiety.

Application

C16 Lactosyl(β) Ceramide (d18:1/16:0) or D-lactosyl-β-1,1′ N-palmitoyl-D-erythro-sphingosine has been used as a lipid standard in mass spectrometry imaging of lipids in human skin disease model hidradenitis suppurativa. It may be used in the preparation of liposomes.

Biochem/physiol Actions

Lactosylceramide (LacCer) is a structural and signaling lipid. It is crucial for signal transduction and cell adhesion. LacCer facilitates microbial attachment and also acts as an adhesion receptor for pathogens. It regulates cellular stress response and apoptosis. LacCer also modulates in cell proliferation and microbial pathogenesis. It is implicated in innate immune responses, host-tissue pathogen interactions and chronic inflammatory diseases.

Packaging

5 mL Amber Glass Screw Cap Vial (860576P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biserka Bakrac et al.
The Journal of biological chemistry, 283(27), 18665-18677 (2008-04-30)
Sphingomyelin (SM) is abundant in the outer leaflet of the cell plasma membrane, with the ability to concentrate in so-called lipid rafts. These specialized cholesterol-rich microdomains not only are associated with many physiological processes but also are exploited as cell
Chemistry, Biochemistry, and Biology of 1-3 Beta Glucans and Related Polysaccharide, 191, 106-114 (2009)
Daniela Couto et al.
Chemistry and physics of lipids, 191, 106-114 (2015-09-01)
Galactosylceramide (GalCer) and lactosylceramide (LacCer) are structural and signaling lipids, playing important roles in signal transduction and cell adhesion. They are especially abundant in the nervous system and in important components of the myelin sheath. Although neurodegenerative disorders are associated
Jarod A Fincher et al.
Scientific reports, 9(1), 17508-17508 (2019-11-27)
Neutral lipids have been implicated in a host of potentially debilitating human diseases, such as heart disease, type-2 diabetes, and metabolic syndrome. Matrix-assisted laser desorption ionization (MALDI), the method-of-choice for mass spectrometry imaging (MSI), has led to remarkable success in
Jamie L Betker et al.
Journal of pharmaceutical sciences (2020-02-01)
Nanoparticle-mediated drug delivery has long utilized PEGylation as a mechanism for reducing uptake by the reticuloendothelial system and extending circulation lifetimes. However, studies over the past 2 decades have established that immune responses to PEG can promote clearance on repeat injection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service