Skip to Content
Merck
All Photos(1)

Key Documents

860526P

Avanti

1-O-Acyl-Ceramide

Avanti Research - A Croda Brand 860526P, powder

Synonym(s):

1-oleoyl-N-heptadecanoyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H101NO4
CAS Number:
Molecular Weight:
816.37
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (860526P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860526P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C53H101NO4/c1-4-7-10-13-16-19-22-25-27-30-33-36-39-42-45-48-53(57)58-49-50(51(55)46-43-40-37-34-31-28-24-21-18-15-12-9-6-3)54-52(56)47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h25,27,43,46,50-51,55H,4-24,26,28-42,44-45,47-49H2,1-3H3,(H,54,56)/b27-25-,

InChI key

FRBOSLBQGQIWQM-YXQBTRHCSA-N

General description

1-O-Acyl-Ceramide is an epidermal ceramide present in mouse and human epidermis. It comprises long chain fatty acids in 1-O-position and is synthesized in the endoplasmic reticulum-related sites.

Application

1-O-Acyl-Ceramide is suitable for use as a lipid standard in
  • high performance thin layer chromatography (HPTLC)
  • in mass and nuclear magnetic resonance spectroscopy for the quantification of lipids isolated from vernix caseosa skin cream
  • in liquid chromatography-tandem mass spectrometry for the quantification of 1-O-Acyl ceramides from liver samples

Biochem/physiol Actions

1-O-Acyl-Ceramide functions in maintaining water barrier homeostasis. Deficiency of the glucosylceramide synthase enzyme leads to the accumulation of O-acylceramides. Treatment of human monoclonal antibody, REMD 2.59 leads to the accumulation of 1-O-acyl-ceramides in soleus muscle and depletion in liver.

Packaging

5 mL Amber Glass Screw Cap Vial (860526P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamoxifen inhibits the biosynthesis of inositolphosphorylceramide in Leishmania
Trinconi CT, et al.
International Journal for Parasitology, Drugs and Drug Resistance, 8(3), 475-487 (2018)
1-O-acylceramides are natural components of human and mouse epidermis
Rabionet M, et al.
Journal of Lipid Research, 54(12), 3312-3321 (2013)
Aline Bayerle et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1865(9), 158741-158741 (2020-06-01)
Except for epidermis and liver, little is known about endogenous expression of 1-O-acylceramides (1-OACs) in mammalian tissue. Therefore, we screened several organs (brain, lung, liver, spleen, lymph nodes, heart, kidney, thymus, small intestine, and colon) from mice for the presence
Glucagon receptor antagonism improves glucose metabolism and cardiac function by promoting AMP-mediated protein kinase in diabetic mice
Sharma AX, et al.
Testing, 22(7), 1760-1773 (2018)
Nonhydroxylated 1-O-acylceramides in vernix caseosa
Harazim E, et al.
Journal of Lipid Research, 59(11), 2164-2173 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service