Skip to Content
Merck
All Photos(2)

Key Documents

860345C

Avanti

14:0 PC-d54

1,2-dimyristoyl-d54-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

DMPC-D54

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H18NO8PD54
CAS Number:
Molecular Weight:
732.27
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (860345C-10mg)
pkg of 1 × 4 mL (860345C-100mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860345C

concentration

10 mg/mL (860345C-10mg)
25 mg/mL (860345C-100mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=O)COC(C([2H])([2H])C([2H])([2H])C([2H])([2H

InChI

1S/C36H72NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-35(38)42-32-34(33-44-46(40,41)43-31-30-37(3,4)5)45-36(39)29-27-25-23-21-19-17-15-13-11-9-7-2/h34H,6-33H2,1-5H3/t34-/m1/s1/i1D3,2D3,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2,19D2,20D2,21D2,22D2,23D2,24D2,25D2,26D2,27D2,28D2,29D2

InChI key

CITHEXJVPOWHKC-RPLUSTTMSA-N

General description

Deuterated fatty acids experience exchange of the deuteriums on the alpha carbon to the carbonyl, i.e., C2 position, and will therefore be a mixture of compounds that are fully deuterated and partially deuterated at that position.

Application

14:0 PC-d54 is suitable to reconstitute [ILV-13CH3, U-15N, 2H] labeled EmrE in n-dodecyl-β-D-maltopyranoside (DDM) back-exchanged to 1H at the amide positions during nuclear magnetic resonance (NMR) sample preparation. It is also suitable for the preparation of phospholipid liposomes.

Packaging

5 mL Clear Glass Sealed Ampule (860345C-100mg)
5 mL Clear Glass Sealed Ampule (860345C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jess Li et al.
The Journal of biological chemistry, 295(9), 2664-2675 (2020-01-25)
Engineering and bioconjugation of proteins is a critically valuable tool that can facilitate a wide range of biophysical and structural studies. The ability to orthogonally tag or label a domain within a multidomain protein may be complicated by undesirable side
Alessio Ausili et al.
Langmuir : the ACS journal of surfaces and colloids, 36(4), 1062-1073 (2020-01-14)
Vitamin K1 and vitamin K2 play very important biological roles as members of chains of electron transport as antioxidants in membranes and as cofactors for the posttranslational modification of proteins that participate in a number of physiological functions such as
Fusion of Legionella pneumophila outer membrane vesicles with eukaryotic membrane systems is a mechanism to deliver pathogen factors to host cell membranes
Jager J, et al.
Cellular Microbiology, 17(5), 607-620 (2015)
Protonation of a glutamate residue modulates the dynamics of the drug transporter EmrE
Gayen A, et al.
Nature chemical biology, 12(3), 141-141 (2016)
Stephen C L Hall et al.
Journal of colloid and interface science, 574, 272-284 (2020-04-25)
Over recent years, there has been a rapid development of membrane-mimetic systems to encapsulate and stabilize planar segments of phospholipid bilayers in solution. One such system has been the use of amphipathic copolymers to solubilize lipid bilayers into nanodiscs. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service