Skip to Content
Merck
All Photos(1)

Key Documents

850811C

Avanti

16:0-22:6 PC-d9

Avanti Research - A Croda Brand

Synonym(s):

1-palmitoyl-2-docosahexaenoyl-sn-glycero-3-phosphocholine-d9

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H71D9NO8P
Molecular Weight:
815.16
UNSPSC Code:
12352211
NACRES:
NA.75

Assay

>99% (TLC)

form

chloroform solution

manufacturer/tradename

Avanti Research - A Croda Brand

packaging

pkg of (pkg of 1 x 1mg (850810C-1MG))

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])[N+](CCOP([O-])(=O)OC[C@@H](COC(=O)CCCCCCCCCCCCCCC)OC(=O)CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CC)(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI key

IESVDEZGAHUQJU-JHLIDBIUSA-N

Packaging

5 mL Clear Glass Sealed Ampule (850811C-1MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service