Skip to Content
Merck
All Photos(2)

Key Documents

850756C

Avanti

16:0-18:2 PE

1-palmitoyl-2-linoleoyl-sn-glycero-3-phosphoethanolamine, chloroform

Synonym(s):

1-hexadecanoyl-2-(9Z,12Z-octadecadienoyl)-sn-glycero-3-phosphoethanolamine; PE(16:0/18:2(9Z,12Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H74NO8P
CAS Number:
Molecular Weight:
715.98
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850756C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850756C

concentration

10 mg/mL (850756C-25mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C39H72NO8P/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-39(42)48-37(36-47-49(43,44)46-34-33-40)35-45-38(41)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h5,7,11,13,17-18,37H,3-4,6,8-10,12,14-16,19-36,40H2,1-2H3,(H,43,44)

InChI key

QONZHKTZKTUIMU-UHFFFAOYSA-N

Application

16:0-18:2 PE has been used for the preparation of lipid hydroperoxide (LOOH) and used in RB-catalyzed photo-, UV photo-, or LOX-1-catalyzed oxidation. It has also been used as a component of liposomes.

Packaging

5 mL Clear Glass Sealed Ampule (850756C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of pure lipid hydroperoxides
Ibusuki D, et al.
Journal of Lipid Research, 49(12), 2668-2677 (2008)
Positional specificity of lysosomal phospholipase A2
Abe A, et al.
Journal of Lipid Research, 47(10), 2268-2279 (2006)
Alwena H Morgan et al.
Nature protocols, 5(12), 1919-1931 (2010-12-04)
Phospholipid-esterified oxylipins include newly described families of bioactive lipids generated by lipoxygenases in immune cells. Until now, assays for their quantitation were not well developed or widely available. Here, we describe a mass spectrometric protocol that enables accurate measurement of
Elitza S Sevova et al.
The Journal of biological chemistry, 285(27), 20580-20587 (2010-05-12)
The Trypanosoma brucei genome has four highly similar genes encoding sphingolipid synthases (TbSLS1-4). TbSLSs are polytopic membrane proteins that are essential for viability of the pathogenic bloodstream stage of this human protozoan parasite and, consequently, can be considered as potential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service