Skip to Content
Merck
All Photos(2)

Documents

850406P

Avanti

4ME 16:0 PA

1,2-diphytanoyl-sn-glycero-3-phosphate (sodium salt), powder

Synonym(s):

1,2-di-(3,7,11,15-tetramethylhexadecanoyl)-sn-glycero-3-phosphate (sodium salt); PA(16:0(3me,7me,11me,15me)/16:0(3me,7me,11me,15me))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H84O8PNa
CAS Number:
Molecular Weight:
783.09
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (850406P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850406P

lipid type

omega FAs

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O)=O)(OC(CC(C)CCCC(C)CCCC(C)CCCC(C)C)=O)COC(CC(C)CCCC(C)CCCC(C)CCCC(C)C)=O.[Na+]

General description

4ME 16:0 PA (1,2-diphytanoyl-sn-glycero-3-phosphate is a fatty acid modified lipid.

Application

4ME 16:0 PA (1,2-diphytanoyl-sn-glycero-3-phosphate (sodium salt)) may be used:
  • as a phospholipid standard to quantify phosphatidic acid in plant samples using mass spectrometry analysis
  • as a component in symmetric/ asymmetric lipid bilayer for biophysical studies
  • to prepare lipid bilayers

Biochem/physiol Actions

Phosphatidic acid (PA) may regulate phagocyte respiratory burst.

Packaging

5 mL Clear Glass Sealed Ampule (850406P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glycerolipids in signal transduction
New comprehensive biochemistry, 31(10), 237-255 null
Extraction and profiling of plant polar glycerol lipids
Liu Y and Wang X
Bio-protocol, 6, 1849e-1849e null
Transient domains of ordered water induced by divalent ions lead to lipid membrane curvature fluctuations
Tarun OB, et al.
Communications Chemistry, 3(1), 1-8 null
Elizabeth A Manrao et al.
PloS one, 6(10), e25723-e25723 (2011-10-13)
Nanopore sequencing has the potential to become a fast and low-cost DNA sequencing platform. An ionic current passing through a small pore would directly map the sequence of single stranded DNA (ssDNA) driven through the constriction. The pore protein, MspA
Jianjun Pan et al.
The journal of physical chemistry. B, 116(51), 14829-14838 (2012-12-04)
Cholesterol and ether lipids are ubiquitous in mammalian cell membranes, and their interactions are crucial in ether lipid mediated cholesterol trafficking. We report on cholesterol's molecular interactions with ether lipids as determined using a combination of small-angle neutron and X-ray

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service