Skip to Content
Merck
All Photos(2)

Key Documents

850350P

Avanti

15:0 PC

1,2-dipentadecanoyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

PC(15:0/15:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H76NO8P
CAS Number:
Molecular Weight:
705.99
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 200 mg (850350P-200mg)
pkg of 1 × 25 mg (850350P-25mg)
pkg of 1 × 500 mg (850350P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850350P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCC)=O)=O

InChI

1S/C38H76NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-37(40)44-34-36(35-46-48(42,43)45-33-32-39(3,4)5)47-38(41)31-29-27-25-23-21-19-17-15-13-11-9-7-2/h36H,6-35H2,1-5H3/p+1

InChI key

LJARBVLDSOWRJT-UHFFFAOYSA-O

General description

Phosphatidylcholines (PCs) are amphipathic molecules. PC accounts for approximately 40–50% of the total cellular phospholipids. They are usually located on the outer leaflets of the biological membrane bilayer.
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

15:0 PC (1,2-dipentadecanoyl-sn-glycero-3-phosphocholine) may be used to prepare giant unilamellar lipid vesicles (GUVs). It has been used as an internal standard for lipidomics analysis in BXD liver samples and plasma samples.

Biochem/physiol Actions

Phosphatidylcholine (PC) may play a vital role in the signaling of neuronal differentiation.

Packaging

20 mL Clear Glass Screw Cap Vial (850350P-500mg)
5 mL Amber Glass Screw Cap Vial (850350P-200mg)
5 mL Amber Glass Screw Cap Vial (850350P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M U Rasheed et al.
The Science of the total environment, 733, 139110-139110 (2020-05-25)
Increased soil nitrogen (N), warming and bark herbivory all are expected to affect boreal forests in the future. We studied the effects of warming (0.5 °C and 4.0 °C above ambient air and soil temperature, respectively), moderate N addition (30 kg N ha-1 y-1) and bark
Thomas Andersen et al.
Soft matter, 10(24), 4268-4274 (2014-05-02)
We reveal that the gel to fluid phase transition causes spherical membrane vesicles to release a finite number of molecules in several consecutive and localized events. By locally melting Giant Unilamellar lipid Vesicles (GUVs), using an optically trapped gold nanoparticle
Zhen Chen et al.
Theriogenology, 144, 56-66 (2020-01-10)
A comparative lipidomic profiling analysis of dairy cattle oocytes with different developmental competences was performed using a combination of high performance liquid chromatography-high resolution tandem mass spectrometry and multivariate statistical analysis. Significant lipidomic changes were identified in degenerating oocytes. Total
Pooja Jha et al.
Cell systems, 6(6), 722-733 (2018-06-18)
The genetics of individual lipid species and their relevance in disease is largely unresolved. We profiled a subset of storage, signaling, membrane, and mitochondrial liver lipids across 385 mice from 47 strains of the BXD mouse population fed chow or
Yasuhiro Horibata et al.
Scientific reports, 10(1), 2845-2845 (2020-02-20)
StarD7 is a phosphatidylcholine (PC)-specific lipid transfer protein essential for the maintenance of mitochondrial PC composition, morphogenesis, and respiration. Here, we studied the role of StarD7 in skeletal myoblast differentiation using mouse myoblast C2C12 cells and human primary myoblasts. Immunofluorescence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service