Skip to Content
Merck
All Photos(2)

Key Documents

840512P

Avanti

16:0 Phosphatidylethanol

1,2-dipalmitoyl-sn-glycero-3-phosphoethanol (sodium salt), powder

Synonym(s):

1,2-dihexadecanoyl-sn-glycero-3-phosphoethanol (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H72O8PNa
CAS Number:
Molecular Weight:
698.93
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (840512P-25mg)
pkg of 1 × 500 mg (840512P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840512P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C37H73O8P.Na/c1-4-7-9-11-13-15-17-19-21-23-25-27-29-31-36(38)42-33-35(34-44-46(40,41)43-6-3)45-37(39)32-30-28-26-24-22-20-18-16-14-12-10-8-5-2;/h35H,4-34H2,1-3H3,(H,40,41);/q;+1/p-1/t35-;/m1./s1

InChI key

MAGNEPCJIVJZIK-RUQJKXHKSA-M

General description

Phosphatidylethanol (PEth) is a phospholipid, that is generated in the the presence of ethanol in cell membranes. PEths like PEth 16:0/18:1, PEth 18:1/18:1, and PEth 16:0/16:0 possesses a common phosphoethanol head. Two fatty acid chains of variable length and degree of saturation are bound to this head group.

Application

16:1 Phosphatidylethanol is suitable to use as a standard for the quantification of PEths in whole blood and in venous or capillary dried blood spots (DBS).

Biochem/physiol Actions

Phosphatidylethanols (PEths) serves as crucial biomarkers of alcohol consumption.

Packaging

20 mL Clear Glass Screw Cap Vial (840512P-500mg)
5 mL Amber Glass Screw Cap Vial (840512P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of phosphatidylethanol 16: 0/18: 1, 18: 1/18: 1, and 16: 0/16: 0 in venous blood and venous and capillary dried blood spots from patients in alcohol withdrawal and control volunteers
Kummer N, et al.
Analytical and Bioanalytical Chemistry, 408(3), 825-838 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service