Skip to Content
Merck
All Photos(3)

Key Documents

840053C

Avanti

Brain PC

Avanti Research - A Croda Brand 840053C

Synonym(s):

L-α-phosphatidylcholine (Brain, Porcine)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840053C-25mg)
pkg of 2 × 4 mL (840053C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840053C

concentration

10 mg/mL (840053C-25mg)
25 mg/mL (840053C-200mg)

lipid type

phosphoglycerides
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)=O

InChI

1S/C42H82NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h20-21,40H,6-19,22-39H2,1-5H3/b21-20-/t40-/m1/s1

InChI key

WTJKGGKOPKCXLL-VYOBOKEXSA-N

General description

Phosphatidylcholine (PC) is a predominant glycerophospholipid in eukaryotes. Brain PC predominantly contains PC 16:0/16:0 and PC 16:0/18:1.

Application

Brain PC is suitable for use in the preparation of lipid mixture for microplate (enzyme linked immunosorbent assay (ELISA))-based recognition of glycolipid-protein interaction assay and liposomes for co-sedimentation assay. Brain PC has been used in the preparation of liposomes for integration of receptors.

Biochem/physiol Actions

Phosphatidylcholine (PC) is the major source of second messengers and lipid mediators. It is intermediate in the synthesis of other phospholipids.

Packaging

5 mL Clear Glass Sealed Ampule (840053C-200mg)
5 mL Clear Glass Sealed Ampule (840053C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphatidylethanolamine and phosphatidylcholine biosynthesis by the Kennedy pathway occurs at different sites in Trypanosoma brucei.
Farine L, et al.
Scientific Reports, 5(5), 16787-16787 (2015)
Choline, Phospholipids, Health, and Disease (1998)
Nanoscale Mobility of the Apo State and TARP Stoichiometry Dictate the Gating Behavior of Alternatively Spliced AMPA Receptors.
Dawe GB, et al.
Neuron, 102(5), 976-992 (2019)
Distinct functions of ATG16L1 isoforms in membrane binding and LC3B lipidation in autophagy-related processes.
Lystad A, et al.
Nature Cell Biology, 21(3), 372-372 (2019)
Geoffrey P Noble et al.
Biochemistry, 54(5), 1180-1187 (2015-01-15)
Misfolding of the prion protein (PrP) plays a central role in the pathogenesis of infectious, sporadic, and inherited prion diseases. Here we use a chemically defined prion propagation system to study misfolding of the pathogenic PrP mutant D177N in vitro.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service