Skip to Content
Merck
All Photos(2)

Key Documents

810195C

Avanti

18:1-12:0 NBD PS

Avanti Research - A Croda Brand 810195C

Synonym(s):

1-oleoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-phosphoserine (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H73N6O13P
CAS Number:
Molecular Weight:
901.04
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810195C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810195C

concentration

1 mg/mL (810195C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylserine (PS) is a minor membrane phospholipid. This anionic phospholipid contains a polar head group and two long hydrophobic acyl chains. 1 7-nitrobenz-2-oxa-1,3-diazol-4-yl (NBD) PS is a fluorescent probe containing NBD at the headgroup of PS.

Application

18:1-12:0 NBD PS or 1-oleoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-phosphoserine (ammonium salt) has been used in donor liposomes to determine the phospholipid transfer activity of the synaptic vesicle membrane protein, VAT-1 and phosphatidic acid (PA) transfer by Ups1−Mdm35 system in mitochondria. 18:1-12:0 NBD PS is also suitable for the determination of in vitro phosphatidylserine decarboxylase (Psd1) activity.

Packaging

5 mL Amber Glass Screw Cap Vial (810195C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organization and dynamics of N-(7-nitrobenz-2-oxa-1, 3-diazol-4-yl)-labeled lipids: a fluorescence approach
Mukherjee S, et al.
Chemistry and Physics of Lipids null
Lipid Homeostasis Is Maintained by Dual Targeting of the Mitochondrial PE Biosynthesis Enzyme to the ER
Jonathan R, et al.
Developmental Cell null
Structural basis for interorganelle phospholipid transport mediated by VAT-1
Watanabe Y, et al.
Journal of biological and chemical chronicles null
Structural and mechanistic insights into phospholipid transfer by Ups1?Mdm35 in mitochondria
Yasunori W, et al.
Nature Communications null
The distribution and function of phosphatidylserine in cellular membranes
Leventis PA and Grinstein S
Annual Review of Biophysics null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service