Skip to Content
Merck
All Photos(2)

Key Documents

810156P

Avanti

18:1-12:0 NBD PE

Avanti Research - A Croda Brand 810156P, powder

Synonym(s):

1-oleoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H70N5O11P
CAS Number:
Molecular Weight:
840.00
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810156P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810156P

shipped in

dry ice

storage temp.

−20°C

General description

N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl) phosphoethanolamine (NBD PE) is a fluorescently labeled lipid where NBD is attached to the headgroup of phosphatidylethanolamine (PE). PE is an aminophospholipid abundant in brain.

Application

18:1-06:0 NBD PE or 1-oleoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-phosphoethanolamine may be used as a component of liposome in cross-linking experiment and to determine M13 protein coat towards phospholipids.

Biochem/physiol Actions

N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl) phosphoethanolamine (NBD PE) plays a key role as a fluorescent analog in the studies of organization and dynamics of membranes. NBD PE contributes to monitoring Red edge excitation shift (REES) effects, with the unique motional and dielectric properties it possesses.

Packaging

5 mL Amber Glass Screw Cap Vial (810156P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fábio Fernandes et al.
Biophysical journal, 87(1), 344-352 (2004-07-09)
Quantification of lipid selectivity by membrane proteins has been previously addressed mainly from electron spin resonance studies. We present here a new methodology for quantification of protein-lipid selectivity based on fluorescence resonance energy transfer. A mutant of M13 major coat
Cardiolipin and phosphatidylethanolamine role in dibucaine interaction with the mitochondrial membrane
Lopes SC, et al.
Biochimica et Biophysica Acta - Biomembranes, 1861(6), 1152-1161 (2019)
Fluorescent methods to study biological membranes, 37-50 (2012)
Quantification of Protein-Lipid Selectivity using FRET: Application to the M13 Major Coat Protein
Fabio F, et al.
Journal of biophysics (Hindawi Publishing Corporation : Online), 87(1), 344-352 (2004)
Organization and dynamics of N-(7-nitrobenz-2-oxa-1, 3-diazol-4-yl)-labeled lipids: a fluorescence approach
Mukherjee S, et al.
Chemistry and Physics of Lipids, 127(1), 91-101 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service