Skip to Content
Merck
All Photos(2)

Key Documents

810146C

Avanti

Egg Liss Rhod PE

Avanti Research - A Croda Brand 810146C

Synonym(s):

L-α-Phosphatidylethanolamine-N-(lissamine rhodamine B sulfonyl) (Ammonium Salt) (Egg-Transphosphatidylated, Chicken)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H107N4O14PS2
CAS Number:
Molecular Weight:
1275.68
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810146C-1mg)
pkg of 5 × 1 mL (810146C-5mg)
pkg of 5 × 2 mL (810146C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810146C

concentration

1 mg/mL (810146C-10mg)
1 mg/mL (810146C-1mg)
1 mg/mL (810146C-5mg)

shipped in

dry ice

storage temp.

−20°C

General description

L-α-Phosphatidylethanolamine (PE) is the second most abundant phospholipid in animals, plants and yeast.

Application

Egg Liss Rhod PE is suitable for the preparation small unilamellar vesicles.

Biochem/physiol Actions

Egg Liss Rhod PE is a fluorescent labelling lipid, useful to study the nanoparticle - cell interactions. L-α-Phosphatidylethanolamine maintains maembrane integrity.

Packaging

5 mL Amber Glass Screw Cap Vial (810146C-10mg)
5 mL Amber Glass Screw Cap Vial (810146C-1mg)
5 mL Amber Glass Screw Cap Vial (810146C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Septins Recognize and Entrap Dividing Bacterial Cells for Delivery to Lysosomes
Krokowski S, et al.
Cell host & microbe, 24(6), 866-874 (2018)
Biodegradable nanoparticles designed for drug delivery: the number of nanoparticles impacts on cytotoxicity
Mendes LP, et al.
Toxicology in vitro, 29(6), 1268-1274 (2015)
Nicolas Puff et al.
Langmuir : the ACS journal of surfaces and colloids, 36(16), 4347-4356 (2020-04-03)
The study of biomimetic model membrane systems undergoing liquid-ordered (Lo)-liquid-disordered (Ld) phase separation using spectroscopic methods has played an important role in understanding the properties of lipid rafts in plasma membranes. In particular, the membrane-associated fluorescence probe Laurdan has proved
Yan Li et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 108(5), 2167-2179 (2020-01-07)
Numerous bioactive molecules produced in cells are involved in the process of bone formation. We consider that appropriate, simultaneous application of two types of bioactive molecules would accelerate the regeneration of tissues and organs. Therefore, we combined aspirin-loaded liposomes (Asp@Lipo)
Sagar Rayamajhi et al.
Colloids and surfaces. B, Biointerfaces, 188, 110804-110804 (2020-01-24)
Endosomal degradation of the nanoparticle is one of the major biological barriers associated with the drug delivery system. Nanoparticles are internalized in the cell via different endocytosis pathways, where they are first delivered to early endosomes which mature to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service