Skip to Content
Merck
All Photos(1)

Documents

800816P

Avanti

16:0 DG

1,2-dipalmitoyl-sn-glycerol, powder

Synonym(s):

1,2-dihexadecanoyl-sn-glycerol; DG(16:0/16:0/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H68O5
CAS Number:
Molecular Weight:
568.91
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (800816P-10mg)
pkg of 1 × 200 mg (800816P-200mg)
pkg of 1 × 25 mg (800816P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 800816P

lipid type

neutral lipids
neutral glycerides

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C35H68O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-34(37)39-32-33(31-36)40-35(38)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h33,36H,3-32H2,1-2H3

InChI key

JEJLGIQLPYYGEE-UHFFFAOYSA-N

General description

16:0 DG (1,2-dipalmitoyl-sn-glycerol) is a longer acyl chains containing diacylglycerol.
In biochemical signaling, diacylglycerol (DAG) functions as a second messenger signaling lipid, and is a product of the hydrolysis of the phospholipid PIP2 (phosphatidylinositolbisphosphate) by the enzyme phospholipase C (PLC) (a membrane-bound enzyme) that, through the same reaction, produces inositol trisphosphate (IP3). Although inositol trisphosphate (IP3) diffuses into the cytosol, DAG remains within the plasma membrane due to its hydrophobic properties. IP3 stimulates the release of calcium ions from the smooth endoplasmic reticulum, whereas DAG is a physiological activator of protein kinase C (PKC). The production of DAG in the membrane facilitates translocation of PKC from the cytosol to the plasma membrane.
Diacylglycerol mimicks the effects of the tumor-promoting compounds phorbol esters.

Application

16:0 DG may be used :
  • to spike brain samples for mass spectrometric analysis
  • as a diacyl-glycerol source in diacylglycerol O-acyltransferase 1 (DGAT1) enzyme assay in human leukemic K562 cells
  • in Golgi-like liposomes reconstitution

Biochem/physiol Actions

16:0 DG (1,2-dipalmitoyl-sn-glycerol) fails to prevent the late fission events in Golgi membrane. Supplementation of 16:0 DG promotes bacterial growth by preventing sporangia.

Packaging

5 mL Amber Glass Screw Cap Vial (800816P-10mg)
5 mL Amber Glass Screw Cap Vial (800816P-200mg)
5 mL Amber Glass Screw Cap Vial (800816P-25mg)

Storage and Stability

Diacylglycerols are conveniently stored in chloroform solutions in glass vials with PTFE-lined caps at -20°C. Under these conditions acyl migration is minimal. Avoid plastic when handling chloroform solutions.

Other Notes

Delivery to cells:
Dry samples of diacylglycerol in chloroform, using a stream of nitrogen. Dissolve the residue in an appropriate volume of ethanol or DMSO, then dilute to the desired aqueous medium.
Effective concentration:
Most biological responses saturate at 20 to 250 μM sn-1,2-dioctanoylglycerol. Only sn-1,2 isomers appear to be active.
Precaution: Since short chain Diacylglycerols mimic effects of the tumor-promoting phorbol diesters in a number of biological systems, extra care should be employed in their handling. Treatment of solutions, vessels and other articles with 1N NaOH before washing or discarding will destroy diacylglycerols.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Probing the global cellular responses to lipotoxicity caused by saturated fatty acids
Piccolis M, et al.
Molecular Cell, 74(1), 32-44 (2019)
1, 2-Dipalmitoyl phosphatidylcholine, 1, 2-dipalmitoyl phosphatidic acid or 1, 2-dipalmitoyl-sn-glycerol inhibit sporangia formation and promote exponential growth of various Frankia isolates from the Casuarinaceae family
Selim S and ScHwencke J
Soil Biology and Biochemistry, 26(5), 569-575 (1994)
Quantification of signaling lipids by nano-electrospray ionization tandem mass spectrometry (Nano-ESI MS/MS)
Haag M, et al.
Metabolites, 2(1), 57-76 (2012)
PTPsigma inhibitors promote hematopoietic stem cell regeneration
Zhang Y, et al.
Nature Communications, 10(1), 1-15 (2019)
B R Ganong et al.
Proceedings of the National Academy of Sciences of the United States of America, 83(5), 1184-1188 (1986-03-01)
The specificity of protein kinase C activation by sn-1,2-diacylglycerols and analogues was investigated by using a Triton X-100 mixed micellar assay [Hannun, Y. A., Loomis, C. R. & Bell, R. M. (1985) J. Biol. Chem. 260, 10039-10043]. Analogues containing acyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service