Skip to Content
Merck
All Photos(1)

Key Documents

800812P

Avanti

12:0 DG

Avanti Research - A Croda Brand

Synonym(s):

1,2-didodecanoyl-sn-glycerol; DG(12:0/12:0/0:0); 110611

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H52O5
CAS Number:
Molecular Weight:
456.70
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

description

1,2-dilauroyl-sn-glycerol

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (800812P-10mg)
pkg of 1 × 200 mg (800812P-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

neutral glycerides
neutral lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O([C@H](COC(=O)CCCCCCCCCCC)CO)C(=O)CCCCCCCCCCC

InChI

1S/C27H52O5/c1-3-5-7-9-11-13-15-17-19-21-26(29)31-24-25(23-28)32-27(30)22-20-18-16-14-12-10-8-6-4-2/h25,28H,3-24H2,1-2H3/t25-/m0/s1

InChI key

OQQOAWVKVDAJOI-VWLOTQADSA-N

General description

sn-1,2-diacylglycerols comprise short saturated acyl side chains (C4-C10)[1] and 12:0 DG is also called 1,2-dilauroyl-sn-glycerol.

Application

12:0 DG has been used as an internal standard for spiking serum samples for liquid chromatography–mass spectrometry analysis.[2] It may be used as an internal standard for spiking RAW 264.7 cells for mass spectroscopy studies[3] and in reverse phase liquid chromatography coupled mass spectrometer for lipidomics studies of human plasma and serum samples.[4]

Biochem/physiol Actions

1,2-dilauroyl-sn-glycerol is a polymerization initiator used in the synthesis of (R)-3-((2-bromo-2-methylpropanoyl)oxy)propane-1,2-diyl didodecanoate (BMPD).[5] sn-1,2-diacylglycerols are prime regulators of biological functions especially in platelets.[6] They also activate protein kinase C.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (800812P-10mg)
5 mL Amber Glass Screw Cap Vial (800812P-200mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipid Profile Characterization and Lipoprotein Comparison of Extracellular Vesicles from Human Plasma and Serum
Sun Y, et al.
Metabolites, 9(11), 259-259 (2019)
Lipidated polymers for the stabilization of cubosomes: nanostructured drug delivery vehicles
Grace JL, et al.
Chemical Communications (Cambridge, England), 53(76), 10552-10555 (2017)
Structural Properties of 1, 2-Diacyl-sn-glycerol in Bulk and at the Water Interface by Molecular Dynamics
van Buuren AR, et al.
Langmuir, 11(8), 2957-2965 (1995)
Quantification of diacylglycerol species from cellular extracts by electrospray ionization mass spectrometry using a linear regression algorithm
Callender HL, et al.
Analytical Chemistry, 79(1), 263-272 (2007)
Serum lipidome analysis of healthy beagle dogs receiving different diets
Boretti FS, et al.
Metabolomics, 16(1), 1-1 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service