Skip to Content
Merck
All Photos(1)

Key Documents

791640C

Avanti

15:0-18:1-d7-PG

Avanti Research - A Croda Brand 791640C

Synonym(s):

1-pentadecanoyl-2-oleoyl(d7)-sn-glycero-3-[phospho-rac-(1′-glycerol)] (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H67D7NaO10 P
CAS Number:
Molecular Weight:
764.01
MDL number:
UNSPSC Code:
41141825
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (791640C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 791640C

concentration

1 mg/mL (791640C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

15:0-18:1-d7-PG (1-pentadecanoyl-2-oleoyl(d7)-sn-glycero-3-[phospho-rac-(1′-glycerol)] is a deuterated phospholipid. The lipid comprises phosphatidylglycerol attached to pentadecanoic acid and oleic acid. Oleic acid harbors the deuterium ions. Phosphatidylglycerol is anionic phospholipid and is associated with thylakoids, microsomal and mitochondrial membrane.
This deuterium labeled lipid is also available in a formulation of multiple lipid standards. The SPLASH LIPIDOMIX Standard includes all of the major lipid classes at ratios relative to human plasma, allowing one easy internal standard solution to be added to every sample.

Application

15:0-18:1-d7-PG (1-pentadecanoyl-2-oleoyl(d7)-sn-glycero-3-[phospho-rac-(1′-glycerol)] (sodium salt),) has been used:
  • as a component of extraction solvent for lipid isolation from biological samples
  • as an internal standard to spike cell lysate and to quantify lipid species using fourier transform mass spectrometry (FTMS) analysis
  • as an isotopically labeled lipid internal standard for correction of lipids detected in lipid droplets using shotgun mass spectrometry analysis

Biochem/physiol Actions

Phosphatidylglycerol (PG) plays a key role in photosynthesis. Cardiolipin is synthesized from PG. PG mediates many mitochondrial functions.

Packaging

2 mL Amber Glass Sealed Ampule (791640C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
LIPIDOMIX is a trademark of Avanti Polar Lipids, LLC
SPLASH is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Shin-Ya Morita et al.
Scientific reports, 5, 11737-11737 (2015-07-01)
Phosphatidylglycerol (PG) and cardiolipin (CL) are synthesized in mitochondria and regulate numerous biological functions. In this study, a novel fluorometric method was developed for measuring PG and CL using combinations of specific enzymes and Amplex Red. This assay quantified the
Florian Geltinger et al.
GeroScience, 42(1), 19-38 (2019-11-05)
Originally Lipid droplets (LDs) were considered as being droplets for lipid storage only. Increasing evidence, however, demonstrates that LDs fulfill a pleiotropy of additional functions. Among them is the modulation of protein as well as lipid homeostasis. Under unfavorable pro-oxidative
Ya Xie et al.
Food chemistry, 319, 126547-126547 (2020-03-18)
In this research, a novel ultrasound-assisted extraction (USAE) method using a one-phase solvent (isopropanol, IPA) combined with liquid chromatography-electrospray ionization quadrupole time-of-flight mass spectrometry (LC-ESI-QToF-MS) was developed for exhaustive profiling of egg yolk lipids, which are considered to be essential
Tong Liu et al.
Expert review of proteomics, 17(3), 233-242 (2020-04-21)
Background: Colorectal cancer (CRC) represents a third leading cause of cancer-related death worldwide. The reliable diagnostic biomarkers for detecting CRC at early stage is critical for decreasing the mortality.Method: A conjunctive lipidomic approach was employed to investigate the differences in
Chuchu Wang et al.
Nature communications, 11(1), 1531-1531 (2020-03-27)
Vesicle associated membrane protein 2 (VAMP2/synaptobrevin2), a core SNARE protein residing on synaptic vesicles (SVs), forms helix bundles with syntaxin-1 and SNAP25 for the SNARE assembly. Prior to the SNARE assembly, the structure of VAMP2 is unclear. Here, by using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service