Skip to Content
Merck
All Photos(2)

Key Documents

700057P

Avanti

22(S)-hydroxycholesterol

Avanti Research - A Croda Brand

Synonym(s):

5-cholestene-3β,22-diol; 22-hydroxycholest-5-en-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

description

cholest-5-ene-3β,22(S)-diol

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (700057P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]1CC[C@@]2([C@@H]3[C@H]([C@H]4[C@@]([C@H](CC4)[C@@H]([C@@H](O)CCC(C)C)C)(CC3)C)CC=C2C1)C

InChI

1S/C27H46O2/c1-17(2)6-11-25(29)18(3)22-9-10-23-21-8-7-19-16-20(28)12-14-26(19,4)24(21)13-15-27(22,23)5/h7,17-18,20-25,28-29H,6,8-16H2,1-5H3/t18-,20-,21-,22+,23-,24-,25-,26-,27+/m0/s1

InChI key

RZPAXNJLEKLXNO-QUOSNDFLSA-N

General description

22(S)-hydroxycholesterol is a stereo isoform of the 22(R)-hydroxycholesterol.

Biochem/physiol Actions

22(S)-hydroxycholesterol (22(S)-HC) promotes glucose catabolism and uptake. It is regarded as a potential target to treat type 2 diabetes. 22(S)-HC also prevents the accumulation of lipids and lipid synthesis in hepatocytes and myotubes. It shows no interaction with liver X receptor (LXR) like 22(R)-hydroxycholesterol and is not estrogenic.

Packaging

5 mL Amber Glass Screw Cap Vial (700057P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroyoshi Sato et al.
Bioscience, biotechnology, and biochemistry, 68(8), 1790-1793 (2004-08-24)
In order to test the estrogenic activity of sterol oxidation products from cholesterol and phytosterols, an estrogen-dependent gene expression assay was performed in estrogen receptor alpha-stably transformed HeLa cells. The ranking of the estrogenic potency of these compounds was different:
Ying Liu et al.
Journal of lipid research, 59(3), 439-451 (2018-01-05)
Cholesterol 25-hydroxylase (CH25H) catalyzes the production of 25-hydroxycholesterol (25-HC), an oxysterol that can play an important role in different biological processes. However, the mechanisms regulating CH25H expression have not been fully elucidated. In this study, we determined that CH25H is
Nina Pettersen Hessvik et al.
The Journal of steroid biochemistry and molecular biology, 128(3-5), 154-164 (2011-11-05)
The aim of this study was to explore the effects of 22(S)-hydroxycholesterol (22(S)-HC) on lipid and glucose metabolism in human-derived cells from metabolic active tissues. Docking of T0901317 and 22(S)-HC showed that both substances fitted into the ligand binding domain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service