330669
Avanti
DOPE ELISA (Clear)
Avanti Research™ - A Croda Brand
Synonym(s):
DOPE ELISA, DOPE ELISA Snoopers™, Clear, for use in absorbance applications
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.84
Recommended Products
form
liquid
manufacturer/tradename
Avanti Research™ - A Croda Brand
packaging
1 ea of 1 × (ELISA strip packaged in foil pouch)
shipped in
wet ice
storage temp.
2-8°C
General description
ELISA Snoopers™ are 8-well strips pre-coated with a single high-quality lipid species. They can be used to investigate protein-lipid interactions, and characterize and titrate lipid binding proteins. This format provides the following advantages:
The 8-well strips are available in optically clear, white, or black surfaces for use in colormetric, luminescent and fluorescent assays, respectively. Avanti Lipid Snoopers ELISA Strips allow multiple sample conditions to be probed simultaneously in a single plate making them a perfect complement to Avanti Lipid Snoopers Nitrocellulose Strips.
- quantitative lipid-protein binding studies
- assemble a 96-well plate using only the lipids of interest for binding studies
- flexibility - use 8 to 96 wells, and 1 to 12 lipids as needed.
The 8-well strips are available in optically clear, white, or black surfaces for use in colormetric, luminescent and fluorescent assays, respectively. Avanti Lipid Snoopers ELISA Strips allow multiple sample conditions to be probed simultaneously in a single plate making them a perfect complement to Avanti Lipid Snoopers Nitrocellulose Strips.
Packaging
ELISA Strip Packaged in Foil Pouch (330669-1EA)
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Snoopers is a trademark of Avanti Polar Lipids, LLC
related product
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service