Skip to Content
Merck
All Photos(1)

Key Documents

100500P

Avanti

E. coli Extract Total

Avanti Research - A Croda Brand

Synonym(s):

E. coli Total Lipid Extract

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

powder

packaging

pkg of 1 × 1 g (100500P-1g)
pkg of 1 × 100 mg (100500P-100mg)
pkg of 5 × 100 mg (100500P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

lipid extracts

shipped in

dry ice

storage temp.

−20°C

General description

This product is an extract of E. coli B (ATCC 11303) grown in Kornberg Minimal media at 37°C and taken at 3/4 log growth phase. Total E. coli lipid extract is a chloroform:methanol extract of the respective tissue. This extract is partitioned against deionized water and the chloroform phase is concentrated.

Application

E. coli Extract Total has been used in the preparation of unilamellar vesicles[1] in lipid screening experiments of membrane protein green fluorescent protein (GFP) based fusion proteins[2] and in the reconstitution of proteoliposomes.[3]

Biochem/physiol Actions

The E. coli lipid extract is a combination of 16, 17 and 18 carbon containing phospholipids namely, phosphatidylethanolamine, phosphatidylglycerol and cardiolipin. It is used for preparing supported lipid bilayers.[4] The E. coli lipid extract serves as a standard for profiling phospholipids, aids in identification and taxonomic grouping of any bacterial sample.[5]

Packaging

5 mL Clear Glass Sealed Ampule (100500P-100mg)
5 mL Clear Glass Sealed Ampule (100500P-500mg)
60 mL Amber Wide Mouth Screw Cap Glass Bottle (100500P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The extracellular gate shapes the energy profile of an ABC exporter
Hutter CAJ, et al.
Nature Communications, 10(1), 2260-2260 (2019)
An engineered thermal-shift screen reveals specific lipid preferences of eukaryotic and prokaryotic membrane proteins
Nji E, et al.
Nature Communications, 9(1), 4253-4253 (2018)
Membrane insertion of small proteins
Protein Secretion, 39-62 (2010)
Lipid compositions in Escherichia coli and Bacillus subtilis during growth as determined by MALDI-TOF and TOF/TOF mass spectrometry
Gidden J, et al.
International Journal of Mass Spectrometry, 283(1-3), 178-184 (2009)
P H Nguyen et al.
Biochemical pharmacology, 174, 113813-113813 (2020-01-20)
P-glycoprotein (Pgp) is an ATP-dependent efflux transporter and plays a major role in anti-cancer drug resistance by pumping a chemically diverse range of cytotoxic drugs from cancerous tumors. Despite numerous studies with the transporter, the molecular features that drive anti-cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service