Key Documents
Z759309
Sigma-Aldrich
Joint clip, standard taper
joint: 19, blue polyethylene
Synonym(s):
NS joint clamps, NS joint clips, National standard joint clamps, National standard joint clips
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
material
blue polyethylene
Joint
joint: 19
packaging
pkg of 50 ea
Related Categories
General description
Color coded, rigid polyethylene joint clips fit standard taper joints. Will not scratch glassware or corrode; withstands temperatures up to 120 °C.
Choose from one of the most recent versions:
Certificates of Analysis (COA)