Z742453
Mircrotube adapter for Orbi-Shaker™ MP and Incu-Mixer™ MP
holds 20 x 1.5-2 mL
All Photos(1)
About This Item
UNSPSC Code:
41103811
NACRES:
NB.73
Recommended Products
feature
holds 20 x 1.5-2 mL
packaging
pkg of 1
manufacturer/tradename
Benchmark Scientific BT1500-A1520
Related Categories
General description
Microtube Adapter for microplate shakers BT1500 and H6004; shakers will fit max 4 adapters
Legal Information
Incu-Mixer is a trademark of Benchmark Scientific, Inc.
Orbi-Shaker is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)