Z740240
QianCap™ gas purging set for Ace pressure tubes and bottles
QianCap™-15-v2, fits no. 15 internal thread
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Recommended Products
General description
QianCap™ Purging-15v2a can be applied onto bottles and pressure tubes with #15 thread. The set has all items required for immediate use. QianCap purging systems can operate easily, safely, and accurately with a wide array of gases, such as H2, O2, CO2, NH3, N2, Ar, D2, CO, for high reproducibility.
Features and Benefits
- Provided with a blue PTFE face and white silicone backing septum that is disposable.
- Septa is an ideal fit on Q-Tube-12 mL.
- The blue PTFE liner has great chemical resistance.
- Silicon backing has a high resealing ability.
Other Notes
Pressure flasks/bottles not included
Legal Information
QianCap is a trademark of Q Labtech LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)