Z677787
Levgo® smartSpatula® disposable PP laboratory spatula, Look for the V at the neck™
L 210 mm, white, standard, sterile (EtO)
Synonym(s):
disposable spatula, microspatula, spatula
About This Item
Recommended Products
material
polypropylene
sterility
sterile (EtO)
packaging
pack of 100 ea
case of 500 ea
manufacturer/tradename
LevGo Inc 17251
L
210 mm
color
white
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
LevGo spatulas are widely used in commercial, university, diagnostic and pharmaceutical labs worldwide as well as by compounding pharmacists to produce customized medications.
Features and Benefits
- The V design at the neck of the disposable lab spatula is a trademark of LevGo, Inc.
- No Searching, washing or contamination.
- 100% Polypropylene is autoclavable and resistant to dilute acids and bases.
- Polypropylene will not shatter after exposure to liquid nitrogen frozen samples.
- Hollow core uses less plastic than other spatulas and minimizes heat or cold transfer.
- Sustainability – LevGo plants a tree for every case sold.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service