Z672122
Tissue processing/embedding cassettes with lid
pkg of 500 ea
Synonym(s):
histopathology cassettes, tissue embedding, tissue processing equipment
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
packaging
pkg of 500 ea
manufacturer/tradename
Simport M490-2
color
white
General description
Disposable plastic cassettes hold tissue specimens during the embedding process, as well as in a storage file. Molded from a special high density polymer these cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service