Key Documents
Z530573
Sigma-Aldrich
Aldrich® reagent bottle
capacity 125 mL, joint: ST/NS 19/22, amber bottle
Synonym(s):
bottle reagent, reagent bottles
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
amber bottle
glass bottle
glass cap
round bottle
Joint
joint: ST/NS19/22
feature
with cap
packaging
pack of 1 ea
manufacturer/tradename
Sigma-Aldrich
capacity
125 mL
General description
Reagent bottles come with a ground glass hex head stopper. For light-sensitive samples, use the amber coated reagent bottles.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC