Key Documents
Z517453
Sigma-Aldrich
Aldrich® cold-finger condenser
joint: ST/NS 29/32
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Pricing and availability is not currently available.
Joint
joint: ST/NS29/32
Overall H
310 mm
cold-finger L × diam.
250 mm × 80 mm
General description
For reactions using low-boiling solvents.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)