Skip to Content
Merck
All Photos(1)

Key Documents

Z256617

Kontes® Williamson microscale glassware kit

Standard kit A

Synonym(s):

748100-0005

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.43

manufacturer/tradename

Kimble®

Looking for similar products? Visit Product Comparison Guide

General description

Kit contains glassware needed to perform experiments described in the textbook Macroscale and Microscale Organic Experiments. This economical microscale glassware kit has a unique, positive connection without the use of threads or ST joints. Glassware is supplied in a foam-lined, corrugated cardboard Ekono-Case.

Features and Benefits

  • Rugged polyethylene storage case (2 1/2 " H x 13 1/2 " W x 10 " D) with a die cut foam insert
  • Kits fit easily into lab bench drawers
  • These MICROFLEX kits feature patented chaste connectors and specially tooled glassware components, providing great versatility
  • Components for Williamson Improved Standard Kit
  • Glassware is manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements

Legal Information

KIMBLE is a registered trademark of DWK Life Sciences
Kontes is a registered trademark of Kimble Kontes Asset Mgmt. Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service