Z255696
Kontes® NMR tube-sealing manifold
Use with 5mm × 9in. L NMR tubes
Synonym(s):
nmr tube, nmr tubes, tube manifold
All Photos(1)
About This Item
UNSPSC Code:
41121705
NACRES:
SB.32
manufacturer/tradename
Kimble®
Related Categories
General description
Single port design with HI-VAC® control valve and aluminum shield for heat deflection. Permits evacuation and flame tip-off of 5mm diam. NMR tube without additional sealing to other glass apparatus. To use, insert NMR tube through the open top cap, place Viton® O-ring over open end of tube, insert tube into the BEVEL-SEAL™ thread, and tighten cap onto thread to make a vacuum seal. Use with 5mm × 9in. L NMR tubes.
Features and Benefits
- Manufactured from borosilicate glass 3.3
- NMR sealing manifolds utilize KONTES® HI-VAC valves
- NMR manifolds feature a BEVEL-SEAL™ threaded connection and a specially designed cap with an aluminum shield for heat deflection
- All O-Rings supplied are FKM
- Valve plugs are 826501-0008
Legal Information
BEVEL-SEAL is a trademark of Kimble Kontes Asset Mgmt. Inc.
HI-VAC is a registered trademark of Kimble Kontes Asset Mgmt. Inc.
KIMBLE is a registered trademark of DWK Life Sciences
Kontes is a registered trademark of Kimble Kontes Asset Mgmt. Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)