Z188964
Kimwipes® disposable wipers
L × W 15 in. × 17 in.
Synonym(s):
cleaning wipes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41103200
NACRES:
NB.35
packaging
pkg of 140 wipes
manufacturer/tradename
Kimberly Clark 34256
L × W
15 in. × 17 in.
Looking for similar products? Visit Product Comparison Guide
General description
Kimwipes™ disposable wipers are suitable for cleaning surfaces, parts, instruments, laboratory lenses, and medical offices. These wipes efficiently remove liquid, dust, and small particles for effective cleanliness.
Application
Kimwipes® disposable wipers have been used in the Porcine mAb immuno-histochemistry (IHC) staining protocol on FMDV-infected cells to remove the remaining blocking solution from the cells after the blocking step.
Features and Benefits
- Delicate precision wiping for crucial tasks with minimized waste through one-at-a-time dispensing.
- Suitable for cleaning glassware and other instruments including other delicate surfaces.
- Equipped with an anti-static dispensing system that reduces lint and electrostatic discharge.
Legal Information
Kimwipes is a registered trademark of Kimberly-Clark Worldwide, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Michael C Puckette et al.
Journal of immunological methods, 487, 112873-112873 (2020-10-01)
This report covers the methodology for generation of stable heterohybridoma clones producing Foot-and-mouth disease virus (FMDV) reactive porcine monoclonal antibodies (mAbs). Swine received five inoculations of an inactivated O1 Manisa FMDV vaccine prior to the harvest of splenocytes. Due to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service