Skip to Content
Merck
All Photos(2)

Documents

Z153192

Cochranes molecular models

minit, classroom set with 5 texts, biochemistry

Synonym(s):

chemistry model sets, molecular model sets

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
60104506
NACRES:
NB.17

description

Scale: 2 cm = 0.1 nanometer (1 Å), Minit 3 cm = 0.1 nanometer (1 Å), Orbit

feature

no. of atoms 1,370
no. of bonds 1,860

manufacturer/tradename

Cochranes 0078

Looking for similar products? Visit Product Comparison Guide

General description

Make models of amino acids, monosaccharides, glycerol, fatty acids, steroids, purines and pyrimidines, peptides, disaccharides, lipids, nucleosides, nucleotides, proteins, polysaccharides, nucleic acids. This group set can be used to make multiple models of small compounds and large tertiary structures. Includes instruction/activity booklet. Includes 5 instruction/activity booklets.

Scale: 2 cm = 0.1 nanometer (1 A), Minit

ATOMS:
  • 1-prong: Hydrogen, white (300), Oxygen, red (50), Sulfur, yellow (20), Chlorine, green (20)
  • 2-prong linear: Hydrogen, white (100), Oxygen, red (100)
  • 2-prong 100°: Sulfur, yellow (20)
  • 2-prong 110°: Nitrogen, blue (20), Oxygen, red (100)
  • 2-prong 120°: Nitrogen, blue (25)
  • 3-prong 108°, 120°, 132°: Carbon, black (25)
  • 3-prong 108°, 126°, 126°: Carbon, black (25)
  • 3-prong 114°, 123°, 123°: Carbon, black (100), Nitrogen, blue (120)
  • 3-prong 120°: Carbon, black (100), Nitrogen, blue (50)
  • 4-prong tetrahedral: Carbon, black (150), Nitrogen, blue (20), Phosphorous, purple (25), Metal, silver (10)
  • 6-prong octahedral: Iron, grey (10)
  • Peptide link (12)

BONDS:
  • Straw, green, 2.5 cm (320)
  • Straw, green, 2 cm (640)
  • Straw, green, 1.5 cm (720)
  • Straw, white, 3.5 cm (160)
  • Straw, mixed set/26 (5)
  • St v flexible white, 2.5 cm (15)
An extensive model system offering 15 atomic bonding configurations in 12 colors of plastic atoms. The carbon atom is represented in nine configurations: linear, trigonal (in five angle variations), tetrahedral, octahedral, trigonal-bipyramidal. Models come in two sizes: Orbit and Minit. Spare atom centers and bonding tubes are available for all model kits.

Please see Technical Information Bulletin AL-183 for a listing of these items, available on our website at sigma-aldrich.com

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service