Z131245
Nalgene® volumetric flask
volume 50 mL, accuracy: 0.10 mL, translucent polypropylene copolymer (PPCO)
Synonym(s):
nalgene, plastic flask, plastic flasks
About This Item
Recommended Products
material
long
translucent polypropylene copolymer (PPCO)
Joint
joint threaded neck13mm
sterility
non-sterile
feature
accuracy class B (ASTM E288 and ISO Standard 384)
closure type Screw cap
packaging
pack of 1
manufacturer/tradename
Nalgene® 4000-0050
volume measuring range
50 mL, accuracy: 0.10 mL
capacity
50 mL
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)