Skip to Content
Merck
All Photos(1)

Key Documents

Z114014

Sigma-Aldrich

Wheaton glass serum bottle

volume 125 mL, clear, O.D. 20 mm, diam. × H 54 mm × 107 mm

Synonym(s):

Wheaton Serum Vial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.11

material

borosilicate glass bottle

description

USP Type I

feature

autoclavable
closure type crimp top vial

O.D.

20 mm

capacity

125 mL

diam. × H

54 mm × 107 mm

volume

125 mL

color

clear

fitting

opening for 20 mm

Looking for similar products? Visit Product Comparison Guide

General description

It is autoclavable, easy sterilization, is used to contain substances to prevent contamination.

Application

  • 28 Validation of a closed system for dispensing serum eye drops.: This study validates a closed system for dispensing serum eye drops, employing Wheaton glass serum bottles to maintain sterility and prevent contamination, which is critical in ophthalmological applications (Hogg et al., 2022).
  • Optimizing the effect of hydrochar on anaerobic digestion of organic fraction municipal solid waste for biogas and methane production.: The research explores the optimization of hydrochar′s effect on anaerobic digestion, utilizing Wheaton glass serum bottles to conduct controlled experiments, ensuring precise measurement of biogas and methane production (Ghasemzadeh et al., 2022).
  • Technical note: Design, development and validation of an automated gas monitoring equipment for measurement of the dynamics of microbial fermentation.: This technical note describes the development of automated gas monitoring equipment, using Wheaton glass serum bottles to monitor microbial fermentation dynamics accurately, contributing to advancements in bioengineering processes (Rahavi et al., 2022).

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roland Wirth et al.
Frontiers in bioengineering and biotechnology, 8, 557572-557572 (2020-10-20)
Microalgae-based bioenergy production is a promising field with regard to the wide variety of algal species and metabolic potential. The use of liquid wastes as nutrient clearly improves the sustainability of microalgal biofuel production. Microalgae and bacteria have an ecological
Leonard I Wassenaar et al.
Rapid communications in mass spectrometry : RCM, 32(3), 184-194 (2017-11-14)
The nitrogen and oxygen (δ δ NO The Cd-azide method yielded excellent results for routine determination of δ
Mark A Altabet et al.
Rapid communications in mass spectrometry : RCM, 33(15), 1227-1239 (2019-04-14)
The nitrogen and oxygen (δ15 N, δ18 O, and δ17 O values) isotopic compositions of nitrate (NO3- ) are crucial tracers of nutrient nitrogen (N) sources and dynamics in aquatic systems. Current methods such as bacterial denitrification or Cd-azide reduction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service