Skip to Content
Merck
All Photos(1)

Key Documents

WHA67205045

Whatman® Polydisc TF Chemical resistant in-line filter

pore size 0.45 μm

Synonym(s):

Whatman filter, Z744263, in line filter, line filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104922
NACRES:
NB.24

material

PTFE membrane
polypropylene housing

sterility

non-sterile

packaging

pkg of 10 ea

manufacturer/tradename

Whatman 6720-5045
Whatman Article No. 28420214 (US reference)

diam.

50 mm

pore size

0.45 μm

Looking for similar products? Visit Product Comparison Guide

General description

Whatman Polydisc TF In-Line Filters from Cytiva are lightweight devices with high chemical and solvent resistance that are particulraly suitable for protective vents or in-line filtration applications.

Polydisc TF aggressive solvent filter

Filtering organic solvents and aggressive solutions calls for a chemically resistant, durable membrane. Cytiva′s Polydisc TF features a PTFE membrane resistant to such compounds. The 1 µm membrane features a polypropylene prefilter for solvent filtration containing high particulate density in order to extend the useful life of the membrane.

Application

  • Pharmaceutical: vents and in-line applications
  • Biotech: sterile vents and exhausts for growth environments, in-line sterilization of gases
  • Laboratory: clean or sterile gases. Filtration of solvents and reagents. Drying gases.
  • Electronics: photoresists, solvents, gases for research

Features and Benefits

  • Chemically resistant hydrophobic PTFE membrane is well suited as a solvent filter for many different solutions
  • Filter can be autoclaved multiple times without losing integrity
  • Integrity-testable (bubble point or water breakthrough pressure in situ methods)
  • Lightweight to prevent tube collapsing

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service