WHA5401090E
Whatman® Moisture Test Paper
diam. (90 mm), pack of 100 ea
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
40161507
NACRES:
NB.24
Recommended Products
material
borosilicate glass
packaging
pack of 100 ea
manufacturer/tradename
Cytiva
diam.
(90 mm)
General description
Moisture test paper for use when drying samples during moisture assessment.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Akihiko Saito et al.
Veterinary ophthalmology, 24(1), 28-36 (2020-09-23)
To compare the superficial punctate fluorescein staining in dogs with and without aqueous tear deficiency. An eye from each client-owned dogs presented to Triangle Animal Eye Clinic between January and December 2018 underwent tear and ocular surface tests, which included
Ahmed Ibrahim et al.
Journal of equine veterinary science, 97, 103344-103344 (2021-01-23)
The exact contribution of the orbital lacrimal gland (OLG) is unknown and is still a subject of debate and inconsistency in literature. Currently, there are no detailed studies addressing the influence of the OLG removal on tear production in equines.
Jing Yang et al.
Cellular and molecular biology (Noisy-le-Grand, France), 66(1), 60-64 (2020-05-04)
Gestational diabetes mellitus (GDM) refers to diabetes mellitus and impaired glucose tolerance first diagnosed during pregnancy without previous history of the disease. Usually, GDM has a high risk of inducing type 2 diabetes mellitus. The incidence of GDM is increasing
Kelsey M Lopez et al.
The Journal of antimicrobial chemotherapy, 75(12), 3576-3581 (2020-08-25)
Reducing Neisseria gonorrhoeae colonies in the oropharynx is a viable solution to minimize the transmission of this bacterium amongst individuals. A strategy involving the electrostatic interaction between a common antiseptic and a discontinued antibiotic (i.e. octenidine and carbenicillin) was evaluated
Natalie S Chow et al.
Veterinary anaesthesia and analgesia, 47(6), 773-780 (2020-08-31)
To evaluate the sedative effects and pharmacokinetics of detomidine gel administered intravaginally to alpacas in comparison with intravenously (IV) administered detomidine. Randomized, crossover, blinded experiment. A group of six healthy adult female Huacaya alpacas (70.3 ± 7.9 kg). Alpacas were
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service