WHA1703050
Whatman® ashless filter aids
Synonym(s):
Whatman filter, Z740764, filter papers
All Photos(1)
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
Recommended Products
sterility
non-sterile
packaging
pkg of 1 ea
manufacturer/tradename
Whatman 1703-050
Whatman Article No. 28418284 (US reference)
Related Categories
General description
Whatman ashless filter aids enhance filtration speed by coagulating precipitates or suspensions to form a thick retentive “prefilter” layer on top of normal filter paper.
Easily dispersible, the powder is of a purity similar to that of Whatman ashless quantitative papers. It is supplied with a two-ended scoop for measuring 0.5 g or 2.5 g quantities.
Easily dispersible, the powder is of a purity similar to that of Whatman ashless quantitative papers. It is supplied with a two-ended scoop for measuring 0.5 g or 2.5 g quantities.
Features and Benefits
- Easily dispersible, the clippings are of purity similar to that of Whatman ashless quantitative papers.
- Supplied with a two-ended scoop for measuring 0.5 g or 2.5
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)