Skip to Content
Merck
All Photos(1)

Key Documents

W433055

Sigma-Aldrich

4-Hydroxyacetophenone

≥95%, FG

Synonym(s):

4′-Hydroxyacetophenone, 1-(4-Hydroxyphenyl)ethanone, 4-Hydroxyphenylethanone, p-Acetophenol, p-Hydroxyphenyl methyl ketone, Piceol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4COCH3
CAS Number:
Molecular Weight:
136.15
FEMA Number:
4330
Beilstein:
774355
MDL number:
UNSPSC Code:
12164502
Flavis number:
07.243
NACRES:
NA.21
Organoleptic:
balsam; hawthorne; mild; sweet
grade:
FG
food allergen:
no known allergens

grade

FG

Quality Level

reg. compliance

EU Regulation 1333/2008 & 178/2002
FDA 21 CFR 117

Assay

≥95%

bp

147-148 °C/3 mmHg (lit.)

mp

109-111 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

balsam; hawthorne; mild; sweet

SMILES string

CC(=O)c1ccc(O)cc1

InChI

1S/C8H8O2/c1-6(9)7-2-4-8(10)5-3-7/h2-5,10H,1H3

InChI key

TXFPEBPIARQUIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

330.8 °F

Flash Point(C)

166 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service