W399701
2-Methyl-4-oxopentane-2-thiol
≥99%, FG
Synonym(s):
4-Methyl-4-mercaptopentan-2-one
About This Item
Halal
Recommended Products
grade
FG
Halal
Quality Level
Agency
meets purity specifications of JECFA
reg. compliance
EU 1334/2008 & 178/2002
FDA 21 CFR 117
Assay
≥99%
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
black currant; meaty; sulfurous
SMILES string
CC(=O)CC(C)(C)S
InChI
1S/C6H12OS/c1-5(7)4-6(2,3)8/h8H,4H2,1-3H3
InChI key
QRNZMFDCKKEPSX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
129.2 °F
Flash Point(C)
54 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service