Skip to Content
Merck
All Photos(1)

Key Documents

W391018

Sigma-Aldrich

Cyclopentanone

≥99%, FG

Synonym(s):

Adipic ketone, Dumasin, Ketopentamethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H8(=O)
CAS Number:
Molecular Weight:
84.12
FEMA Number:
3910
Beilstein:
605573
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.149
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

Assay

≥99%

refractive index

n20/D 1.437 (lit.)

bp

130-131 °C (lit.)

mp

−51 °C (lit.)

density

0.951 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

minty

SMILES string

O=C1CCCC1

InChI

1S/C5H8O/c6-5-3-1-2-4-5/h1-4H2

InChI key

BGTOWKSIORTVQH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Cyclopentanone is a cyclic ketone that occurs in allium species, beef, cheese, and chicken. It is generally used as a fragrance ingredient in fragrances and in non-cosmetic products.

Application


  • High-oxygen-modified atmospheric packaging delays flavor and quality deterioration in fresh-cut broccoli.: This research investigates the impact of high-oxygen-modified atmospheric packaging on the preservation of fresh-cut broccoli. Cyclopentanone is identified as a significant compound in the aroma profile, contributing to the understanding of packaging technologies that maintain food quality and extend shelf life. (He et al., 2024).

  • Selective production of methylindan and tetralin with xylose or hemicellulose.: The study focuses on the selective production of methylindan and tetralin using xylose or hemicellulose, with Cyclopentanone playing a key role in the catalytic processes. This research provides insights into sustainable chemical production from biomass-derived feedstocks. (Zou et al., 2024).

  • Synthesis of Hydroxylamine via Ketone-Mediated Nitrate Electroreduction.: This paper presents a novel method for synthesizing hydroxylamine through ketone-mediated nitrate electroreduction, utilizing Cyclopentanone. The findings demonstrate the versatility of Cyclopentanone in electrochemical synthesis and its potential for green chemistry applications. (Jia et al., 2024).


Other Notes

Handle and store under inert gas.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

86.0 °F - closed cup

Flash Point(C)

30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fragrance material review on cyclopentanone.
Scognamiglio J, et al.
Food And Chemical Toxicology, 50, S608-S612 (2012)
Mohi Uddin Jewel et al.
Molecules (Basel, Switzerland), 25(5) (2020-03-04)
Fully inkjet-printed device fabrication is a crucial goal to enable large-area printed electronics. The limited number of two-dimensional (2D) material inks, the bottom-gated structures, and the low current on/off ratio of thin-film transistors (TFTs) has impeded the practical applications of
Pragya Verma et al.
The Journal of organic chemistry, 76(14), 5606-5613 (2011-06-02)
The density functional theory investigation on the mechanism of NHC-catalyzed cycloannulation reaction of the homoenolate derived from butenal with pentenone is studied. The M06-2X/6-31+G** and B3LYP/6-31+G** levels of theory, including the effect of continuum solvation in dichloromethane and tetrahydrofuran, are
Na Lin et al.
Physical chemistry chemical physics : PCCP, 14(10), 3669-3680 (2012-02-09)
We present a theoretical study of vibrationally resolved circular dichroism spectra, both in the adiabatic and non-adiabatic frameworks, with a full account of Franck-Condon and Herzberg-Teller vibrational contributions for the former. Model calculations have been performed on 2(R)-deuteriocyclopentanone, whose chirality
Anqi Ma et al.
Organic letters, 12(16), 3634-3637 (2010-08-14)
The O-TMS-protected diphenylprolinol-catalyzed cascade double Michael addition reactions of alpha,beta-unsaturated aldehydes with a beta-keto ester bearing a highly electron-deficient olefin unit occur smoothly to afford polysubstituted cyclopentanones. This process allows formation of four contiguous stereocenters in the cyclopentanone ring in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service