Skip to Content
Merck
All Photos(1)

Key Documents

W383503

Sigma-Aldrich

Ethyl (methylthio)acetate

≥98%, FG

Synonym(s):

(Methylthio)acetic acid ethyl ester, Ethyl α-(methylthio)acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2CO2C2H5
CAS Number:
Molecular Weight:
134.20
FEMA Number:
3835
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.122
NACRES:
NA.21
grade:
FG
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG

Agency

meets requirements for JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥98%

color

colorless

refractive index

n20/D 1.459 (lit.)

bp

70-72 °C/25 mmHg (lit.)

density

1.043 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; tropical; fruity; sulfurous

SMILES string

CCOC(=O)CSC

InChI

1S/C5H10O2S/c1-3-7-5(6)4-8-2/h3-4H2,1-2H3

InChI key

MDIAKIHKBBNYHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Natural occurrence: Apple and melon.

Biochem/physiol Actions

Odor at 1.0%
Taste at 1-2 ppm

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 30, 39-39 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service